missing translation for 'onlineSavingsMsg'
Learn More

2,5-Diiodo-1-methylimidazole, 98%, Thermo Scientific Chemicals

$451.92 - $451.92

Chemical Identifiers

CAS 86026-81-5
Molecular Formula C4H4I2N2
Molecular Weight (g/mol) 333.899
MDL Number MFCD02179540
InChI Key SZYKHAFVWJNSQU-UHFFFAOYSA-N
PubChem CID 335838
IUPAC Name 2,5-diiodo-1-methylimidazole
SMILES CN1C(=CN=C1I)I
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5121203
View Documents
Thermo Scientific Chemicals
H5121203
1 g
Each for $451.92
Only null left
 
Description

Description

It is employed as intermediate for pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as intermediate for pharmaceutical.

Notes
Light Sensitive, store in dark. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications

Chemical Identifiers

86026-81-5
333.899
SZYKHAFVWJNSQU-UHFFFAOYSA-N
2,5-diiodo-1-methylimidazole
C4H4I2N2
MFCD02179540
335838
CN1C(=CN=C1I)I

Specifications

86026-81-5
MFCD02179540
Light sensitive
SZYKHAFVWJNSQU-UHFFFAOYSA-N
2,5-diiodo-1-methylimidazole
335838
98%
C4H4I2N2
1 g
2,5-diiodo-1-methyl-1h-imidazole, pubchem8993, acmc-1bke8, 1-methyl-2,5-diiodoimidazole, 1-methyl-2,5-diiodo-1h-imidazole, 1h-imidazole,2,5-diiodo-1-methyl, 2,5-bis iodanyl-1-methyl-imidazole, 1h-imidazole, 2,5-diiodo-1-methyl, 2,5-diiodo-1-methylimidazole
CN1C(=CN=C1I)I
333.899
333.9
2,5-Diiodo-1-methylimidazole
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only