missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dihydrofuran 98.0+%, TCI America™

$115.57 - $635.65

Chemical Identifiers

CAS 1708-29-8
Molecular Formula C4H6O
Molecular Weight (g/mol) 70.091
MDL Number MFCD00003219
InChI Key ARGCQEVBJHPOGB-UHFFFAOYSA-N
Synonym furan, 2,5-dihydro, 3-oxolene, 1-oxa-3-cyclopentene, 2,5-dihydro-furan, furan,5-dihydro, oxa-3-cyclopentene, 2,5 dihydrofurane, 2,5-dihydro-fura, pubchem3987, acmc-1buc6
PubChem CID 15570
IUPAC Name 2,5-dihydrofuran
SMILES C1C=CCO1
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
D055125ML
View Documents
TCI America
D055125ML
25 mL
Each for $115.57
Only null left
 
D0551100ML
View Documents
TCI America
D0551100ML
100 mL
Each for $255.38
Only null left
 
D0551500ML
View Documents
TCI America
D0551500ML
500 mL
Each for $635.65
Only null left
 
Description

Description

Specifications

Chemical Identifiers

1708-29-8
70.091
ARGCQEVBJHPOGB-UHFFFAOYSA-N
15570
C1C=CCO1
C4H6O
MFCD00003219
furan, 2,5-dihydro, 3-oxolene, 1-oxa-3-cyclopentene, 2,5-dihydro-furan, furan,5-dihydro, oxa-3-cyclopentene, 2,5 dihydrofurane, 2,5-dihydro-fura, pubchem3987, acmc-1buc6
2,5-dihydrofuran

Specifications

1708-29-8
67°C
MFCD00003219
1993
ARGCQEVBJHPOGB-UHFFFAOYSA-N
2,5-dihydrofuran
15570
≥98.0% (GC)
2,5-Dihydrofuran
Yellow
C4H6O
25 mL
furan, 2,5-dihydro, 3-oxolene, 1-oxa-3-cyclopentene, 2,5-dihydro-furan, furan,5-dihydro, oxa-3-cyclopentene, 2,5 dihydrofurane, 2,5-dihydro-fura, pubchem3987, acmc-1buc6
C1C=CCO1
70.091
70.09
Liquid
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (9)-0731

RTECSNumber : LU0660000

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'