missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(4-tert-Butylphenyl)-5-(4-biphenylyl)-1,3,4-oxadiazole (purified by sublimation) 99.0+%, TCI America™
$182.68 - $490.25
Chemical Identifiers
CAS | 15082-28-7 |
---|---|
Molecular Formula | C24H22N2O |
Molecular Weight (g/mol) | 354.453 |
MDL Number | MFCD00003101 |
InChI Key | XZCJVWCMJYNSQO-UHFFFAOYSA-N |
Synonym | butyl pbd, butyl-pbd, 2-4-biphenylyl-5-4-tert-butylphenyl-1,3,4-oxadiazole, 2-4-tert-butylphenyl-5-4-biphenylyl-1,3,4-oxadiazole, 2-1,1'-biphenyl-4-yl-5-4-tert-butyl phenyl-1,3,4-oxadiazole, 2-4-tert-butylphenyl-5-4-biphenyl-1,3,4-oxadiazole, 1,3,4-oxadiazole, 2-1,1'-biphenyl-4-yl-5-4-1,1-dimethylethyl phenyl, 2-biphenyl-4-yl-5-4-tert-butylphenyl-1,3,4-oxadiazole, 1,3,4-oxadiazole, 2-4-biphenylyl-5-p-tert-butylphenyl, 2-t-butylphenyl-5-4-biphenylyl-1,3,4-oxadiazole |
PubChem CID | 84782 |
IUPAC Name | 2-(4-tert-butylphenyl)-5-(4-phenylphenyl)-1,3,4-oxadiazole |
SMILES | CC(C)(C)C1=CC=C(C=C1)C2=NN=C(O2)C3=CC=C(C=C3)C4=CC=CC=C4 |
Chemical Identifiers
15082-28-7 | |
354.453 | |
XZCJVWCMJYNSQO-UHFFFAOYSA-N | |
84782 | |
CC(C)(C)C1=CC=C(C=C1)C2=NN=C(O2)C3=CC=C(C=C3)C4=CC=CC=C4 |
C24H22N2O | |
MFCD00003101 | |
butyl pbd, butyl-pbd, 2-4-biphenylyl-5-4-tert-butylphenyl-1,3,4-oxadiazole, 2-4-tert-butylphenyl-5-4-biphenylyl-1,3,4-oxadiazole, 2-1,1'-biphenyl-4-yl-5-4-tert-butyl phenyl-1,3,4-oxadiazole, 2-4-tert-butylphenyl-5-4-biphenyl-1,3,4-oxadiazole, 1,3,4-oxadiazole, 2-1,1'-biphenyl-4-yl-5-4-1,1-dimethylethyl phenyl, 2-biphenyl-4-yl-5-4-tert-butylphenyl-1,3,4-oxadiazole, 1,3,4-oxadiazole, 2-4-biphenylyl-5-p-tert-butylphenyl, 2-t-butylphenyl-5-4-biphenylyl-1,3,4-oxadiazole | |
2-(4-tert-butylphenyl)-5-(4-phenylphenyl)-1,3,4-oxadiazole |
Specifications
15082-28-7 | |
White | |
MFCD00003101 | |
butyl pbd, butyl-pbd, 2-4-biphenylyl-5-4-tert-butylphenyl-1,3,4-oxadiazole, 2-4-tert-butylphenyl-5-4-biphenylyl-1,3,4-oxadiazole, 2-1,1'-biphenyl-4-yl-5-4-tert-butyl phenyl-1,3,4-oxadiazole, 2-4-tert-butylphenyl-5-4-biphenyl-1,3,4-oxadiazole, 1,3,4-oxadiazole, 2-1,1'-biphenyl-4-yl-5-4-1,1-dimethylethyl phenyl, 2-biphenyl-4-yl-5-4-tert-butylphenyl-1,3,4-oxadiazole, 1,3,4-oxadiazole, 2-4-biphenylyl-5-p-tert-butylphenyl, 2-t-butylphenyl-5-4-biphenylyl-1,3,4-oxadiazole | |
CC(C)(C)C1=CC=C(C=C1)C2=NN=C(O2)C3=CC=C(C=C3)C4=CC=CC=C4 | |
354.453 | |
354.45 | |
Crystalline Powder |
138°C | |
C24H22N2O | |
1 g | |
XZCJVWCMJYNSQO-UHFFFAOYSA-N | |
2-(4-tert-butylphenyl)-5-(4-phenylphenyl)-1,3,4-oxadiazole | |
84782 | |
≥99.0% (GC) | |
2-(4-tert-Butylphenyl)-5-(4-biphenylyl)-1,3,4-oxadiazole (purified by sublimation) |
Safety and Handling
TSCA : Yes