missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dinitrophenyl Thiocyanate 98.0+%, TCI America™

$95.06 - $861.19

Chemical Identifiers

CAS 1594-56-5
Molecular Formula C7H3N3O4S
Molecular Weight (g/mol) 225.178
MDL Number MFCD00047702
InChI Key XQDQRCRASHAZBA-UHFFFAOYSA-N
Synonym dntb, nirit, trirodazeen, gryzbol, grzybol, 2,4-dinitrothiocyanobenzene, nirit supra, tri-rodazene, 2,4-dinitrothiocyanatobenzene, rhodandinitrobenzol
PubChem CID 15325
ChEBI CHEBI:53051
IUPAC Name (2,4-dinitrophenyl) thiocyanate
SMILES C1=CC(=C(C=C1[N+](=O)[O-])[N+](=O)[O-])SC#N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
T01991G
View Documents
TCI America
T01991G
1 g
Each for $95.06
Only null left
 
T019925G
View Documents
TCI America
T019925G
25 g
Each for $861.19
Only null left
 
Description

Description

Specifications

Chemical Identifiers

1594-56-5
225.178
XQDQRCRASHAZBA-UHFFFAOYSA-N
15325
(2,4-dinitrophenyl) thiocyanate
C7H3N3O4S
MFCD00047702
dntb, nirit, trirodazeen, gryzbol, grzybol, 2,4-dinitrothiocyanobenzene, nirit supra, tri-rodazene, 2,4-dinitrothiocyanatobenzene, rhodandinitrobenzol
CHEBI:53051
C1=CC(=C(C=C1[N+](=O)[O-])[N+](=O)[O-])SC#N

Specifications

1594-56-5
Yellow
MFCD00047702
2811
XQDQRCRASHAZBA-UHFFFAOYSA-N
(2,4-dinitrophenyl) thiocyanate
15325
225.18
Crystalline Powder
139°C
C7H3N3O4S
1 g
dntb, nirit, trirodazeen, gryzbol, grzybol, 2,4-dinitrothiocyanobenzene, nirit supra, tri-rodazene, 2,4-dinitrothiocyanatobenzene, rhodandinitrobenzol
C1=CC(=C(C=C1[N+](=O)[O-])[N+](=O)[O-])SC#N
225.178
CHEBI:53051
≥98.0% (GC)
2,4-Dinitrophenyl Thiocyanate
Videos
Safety and Handling

Safety and Handling

RTECSNumber : XK9450000

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'