missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4,6-Tris(2-pyridyl)-s-triazine, For Spectrophotometric Det. of Fe, 98%, MilliporeSigma™ Supelco™
2,4,6-Tris(2-pyridyl)-s-triazine is a complexing reagent used in spectrophotometric analysis.
Supplier: Merck Emd Millipore T125325G
Specifications
3682-35-7 | |
Glass Bottle | |
C18H12N6 | |
25 g | |
KMVWNDHKTPHDMT-UHFFFAOYSA-N | |
tris(pyridin-2-yl)-1,3,5-triazine | |
312.33 | |
For spectrophotometric det. (of Fe) |
247°C to 249°C | |
C18H12N6 | |
MFCD00006045 | |
2,4,6-Tri(2-pyridyl)-s-triazine; TPTZ | |
C1=CC=C(N=C1)C1=NC(=NC(=N1)C1=CC=CC=N1)C1=CC=CC=N1 | |
312.34 | |
≥98% |
Chemical Identifiers
3682-35-7 | |
312.34 | |
KMVWNDHKTPHDMT-UHFFFAOYSA-N | |
tris(pyridin-2-yl)-1,3,5-triazine |
C18H12N6 | |
MFCD00006045 | |
2,4,6-Tri(2-pyridyl)-s-triazine; TPTZ | |
C1=CC=C(N=C1)C1=NC(=NC(=N1)C1=CC=CC=N1)C1=CC=CC=N1 |
Safety and Handling
RTECSNumber : XZ2050000