Learn More
Thermo Scientific Chemicals 2,3:4,6-Di-O-isopropylidene-alpha-L-sorbofuranose, 98%
CAS: 14131-84-1 | C12H20O6 | 260.286 g/mol
Supplier: Thermo Scientific Chemicals J6630309
Description
2,3:5,6-Di-O-isopropylidene-alpha-D-mannofuranose is used in the syntheses of ovalicin1 and of the sugar core of hikizimycin.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2,3:5,6-Di-O-isopropylidene-alpha-D-mannofuranose is used in the syntheses of ovalicin1 and of the sugar core of hikizimycin.
Solubility
Soluble in acetone, methanol, water.
Notes
Hygroscopic. Protect from heat. Store under dry inert gas. Incompatible with moisture and oxidizing agents.
Specifications
2,3:5,6-Di-O-isopropylidene-alpha-D-mannofuranose | |
125°C to 126°C (decomposition) | |
C12H20O6 | |
10 g | |
Hygroscopic | |
Soluble in acetone,methanol,water. | |
CC1(OCC(O1)C2C3C(C(O2)O)OC(O3)(C)C)C | |
260.286 | |
260.28 | |
Powder |
14131-84-1 | |
White | |
MFCD00134206 | |
84382 | |
2,3:5,6-bis-o-1-methylethylidene-, a-d-mannofuranose, 3ar,4s,6r-6-2,2-dimethyl-1,3-dioxolan-4-yl-2,2-dimethyl-tetrahydrofuro 3,4-d 1,3 dioxol-4-ol | |
JWWCLCNPTZHVLF-WVHJSVDISA-N | |
(3aR,4S,6R)-6-(2,2-dimethyl-1,3-dioxolan-4-yl)-2,2-dimethyl-3a,4,6,6a-tetrahydrofuro[3,4-d][1,3]dioxol-4-ol | |
45039045 | |
98% |
Chemical Identifiers
14131-84-1 | |
260.286 | |
JWWCLCNPTZHVLF-WVHJSVDISA-N | |
45039045 | |
CC1(OCC(O1)C2C3C(C(O2)O)OC(O3)(C)C)C |
C12H20O6 | |
MFCD00134206 | |
2,3:5,6-bis-o-1-methylethylidene-, a-d-mannofuranose, 3ar,4s,6r-6-2,2-dimethyl-1,3-dioxolan-4-yl-2,2-dimethyl-tetrahydrofuro 3,4-d 1,3 dioxol-4-ol | |
(3aR,4S,6R)-6-(2,2-dimethyl-1,3-dioxolan-4-yl)-2,2-dimethyl-3a,4,6,6a-tetrahydrofuro[3,4-d][1,3]dioxol-4-ol |
Safety and Handling
TSCA : No
Recommended Storage : Keep cold; Store under Argon
RUO – Research Use Only