missing translation for 'onlineSavingsMsg'
Learn More

2,3:4,5-Di-O-isopropylidene-beta-D-fructopyranose 98.0+%, TCI America™

$54.06 - $113.71

Chemical Identifiers

CAS 20880-92-6
Molecular Formula C12H20O6
Molecular Weight (g/mol) 260.286
MDL Number MFCD00022183
InChI Key PSSHGMIAIUYOJF-OZFQHSNDSA-N
Synonym 2,3:4,5-bis-o-1,2 methylethylidene-beta-d-fruetopyranose
PubChem CID 45357248
IUPAC Name [(3aR,5aS,8aS)-2,2,7,7-tetramethyl-5,5a,8a,8b-tetrahydrodi[1,3]dioxolo[4,5-a
SMILES CC1(OC2COC3(C(C2O1)OC(O3)(C)C)CO)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
D37585G
View Documents
TCI America
D37585G
5 g
Each for $54.06
Only null left
 
D375825G
View Documents
TCI America
D375825G
25 g
Each for $113.71
Only null left
 
Description

Description

Specifications

Chemical Identifiers

20880-92-6
260.286
PSSHGMIAIUYOJF-OZFQHSNDSA-N
45357248
CC1(OC2COC3(C(C2O1)OC(O3)(C)C)CO)C
C12H20O6
MFCD00022183
2,3:4,5-bis-o-1,2 methylethylidene-beta-d-fruetopyranose
[(3aR,5aS,8aS)-2,2,7,7-tetramethyl-5,5a,8a,8b-tetrahydrodi[1,3]dioxolo[4,5-a

Specifications

20880-92-6
White
MFCD00022183
2,3:4,5-bis-o-1,2 methylethylidene-beta-d-fruetopyranose
CC1(OC2COC3(C(C2O1)OC(O3)(C)C)CO)C
260.286
260.29
Crystalline Powder
96°C
C12H20O6
5 g
PSSHGMIAIUYOJF-OZFQHSNDSA-N
[(3aR,5aS,8aS)-2,2,7,7-tetramethyl-5,5a,8a,8b-tetrahydrodi[1,3]dioxolo[4,5-a
45357248
≥98.0% (GC)
2,3:4,5-Di-O-isopropylidene-beta-D-fructopyranose
Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Freezer

SDS
missing translation for 'documents'

missing translation for 'documents'