missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3,3-Trimethyl-3H-benzo[g]indole 98.0+%, TCI America™
$229.29 - $781.03
Chemical Identifiers
CAS | 74470-85-2 |
---|---|
Molecular Formula | C15H15N |
Molecular Weight (g/mol) | 209.292 |
MDL Number | MFCD00723378 |
InChI Key | MNRRNPKQXGBGBH-UHFFFAOYSA-N |
Synonym | 2,3,3-trimethyl-3h-benzo g indole, 2,3,3-trimethylbenzo g indole, 2,3,3-trimethyl-3h-benz g indole, 3h-benz g indole, 2,3,3-trimethyl, 2,3,3-trimethyl-3h-benz g indol, acmc-209ouz, 2,3,3-trimethylbenzindole, 2,3,3-trimethyl-6,7-benzoindolenine, 2,3,3-trimethyl-3-hydrobenzo g indole |
PubChem CID | 12871551 |
IUPAC Name | 2,3,3-trimethylbenzo[g]indole |
SMILES | CC1=NC2=C(C1(C)C)C=CC3=CC=CC=C32 |
Chemical Identifiers
74470-85-2 | |
209.292 | |
MNRRNPKQXGBGBH-UHFFFAOYSA-N | |
12871551 | |
CC1=NC2=C(C1(C)C)C=CC3=CC=CC=C32 |
C15H15N | |
MFCD00723378 | |
2,3,3-trimethyl-3h-benzo g indole, 2,3,3-trimethylbenzo g indole, 2,3,3-trimethyl-3h-benz g indole, 3h-benz g indole, 2,3,3-trimethyl, 2,3,3-trimethyl-3h-benz g indol, acmc-209ouz, 2,3,3-trimethylbenzindole, 2,3,3-trimethyl-6,7-benzoindolenine, 2,3,3-trimethyl-3-hydrobenzo g indole | |
2,3,3-trimethylbenzo[g]indole |
Specifications
74470-85-2 | |
127°C | |
MFCD00723378 | |
2,3,3-trimethyl-3h-benzo g indole, 2,3,3-trimethylbenzo g indole, 2,3,3-trimethyl-3h-benz g indole, 3h-benz g indole, 2,3,3-trimethyl, 2,3,3-trimethyl-3h-benz g indol, acmc-209ouz, 2,3,3-trimethylbenzindole, 2,3,3-trimethyl-6,7-benzoindolenine, 2,3,3-trimethyl-3-hydrobenzo g indole | |
CC1=NC2=C(C1(C)C)C=CC3=CC=CC=C32 | |
209.292 | |
209.29 | |
Crystalline Powder |
78°C | |
C15H15N | |
5 g | |
MNRRNPKQXGBGBH-UHFFFAOYSA-N | |
2,3,3-trimethylbenzo[g]indole | |
12871551 | |
≥98.0% (HPLC,T) | |
2,3,3-Trimethyl-3H-benzo[g]indole |
Safety and Handling
TSCA : No