missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,2'-Thiodiglycolic Acid 98.0+%, TCI America™
$52.19 - $507.02
Chemical Identifiers
CAS | 123-93-3 |
---|---|
Molecular Formula | C4H6O4S |
Molecular Weight (g/mol) | 150.148 |
MDL Number | MFCD00004359 |
InChI Key | UVZICZIVKIMRNE-UHFFFAOYSA-N |
Synonym | thiodiglycolic acid, 2,2'-thiodiacetic acid, thiodiacetic acid, mercaptodiacetic acid, tdga, dicarboxymethyl sulfide, thiodi acetic acid, carboxymethylthio acetic acid, 2,2'-thiodiethanoic acid, thiodiglycollic acid |
PubChem CID | 31277 |
ChEBI | CHEBI:35012 |
IUPAC Name | 2-(carboxymethylsulfanyl)acetic acid |
SMILES | C(C(=O)O)SCC(=O)O |
Chemical Identifiers
123-93-3 | |
150.148 | |
UVZICZIVKIMRNE-UHFFFAOYSA-N | |
31277 | |
2-(carboxymethylsulfanyl)acetic acid |
C4H6O4S | |
MFCD00004359 | |
thiodiglycolic acid, 2,2'-thiodiacetic acid, thiodiacetic acid, mercaptodiacetic acid, tdga, dicarboxymethyl sulfide, thiodi acetic acid, carboxymethylthio acetic acid, 2,2'-thiodiethanoic acid, thiodiglycollic acid | |
CHEBI:35012 | |
C(C(=O)O)SCC(=O)O |
Specifications
130°C | |
White | |
MFCD00004359 | |
thiodiglycolic acid, 2,2'-thiodiacetic acid, thiodiacetic acid, mercaptodiacetic acid, tdga, dicarboxymethyl sulfide, thiodi acetic acid, carboxymethylthio acetic acid, 2,2'-thiodiethanoic acid, thiodiglycollic acid | |
C(C(=O)O)SCC(=O)O | |
150.148 | |
CHEBI:35012 | |
≥98.0% (GC,T) | |
Crystalline Powder |
123-93-3 | |
C4H6O4S | |
3261 | |
UVZICZIVKIMRNE-UHFFFAOYSA-N | |
2-(carboxymethylsulfanyl)acetic acid | |
31277 | |
150.15 | |
25 g | |
2,2′-Thiodiglycolic Acid |
Safety and Handling
RTECSNumber : AJ6475000
TSCA : Yes