missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(2-Phthalimidoethoxy)acetic acid, 97%
CAS: 69676-65-9 | C12H11NO5 | 249.22 g/mol
$105.04 - $353.05
Chemical Identifiers
CAS | 69676-65-9 |
---|---|
Molecular Formula | C12H11NO5 |
Molecular Weight (g/mol) | 249.22 |
MDL Number | MFCD06661364 |
InChI Key | PHZYUQLIZKTSJE-UHFFFAOYSA-N |
Synonym | 2-2-1,3-dioxoisoindolin-2-yl ethoxy acetic acid, 2-2-phthalimidoethoxy acetic acid, 2-2-phthalimidoethoxy-acetic acid, 2-1,3-dioxoisoindol-2-yl ethoxy acetic acid, 2-2-1,3-dioxoisoindol-2-yl ethoxy acetic acid, 2-phthalimido ethoxyacetic acid, 2-1,3-dioxoisoindolin-2-yl ethoxy acetic acid, 2-2-1,3-dioxoisoindolin-2-yl ethoxy aceticacid, 2-1,3-dioxo-1,3-dihydro-2h-isoindol-2-yl ethoxy acetic acid, 2-2-1,3-dioxo-2,3-dihydro-1h-isoindol-2-yl ethoxy acetic acid |
PubChem CID | 2755407 |
IUPAC Name | 2-[2-(1,3-dioxoisoindol-2-yl)ethoxy]acetic acid |
SMILES | OC(=O)COCCN1C(=O)C2=CC=CC=C2C1=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH3398803
|
Thermo Scientific Chemicals
H3398803 |
1 g |
Each for $105.04
|
|
|||||
AAH3398806
|
Thermo Scientific Chemicals
H3398806 |
5 g |
Each for $353.05
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
69676-65-9 | |
249.22 | |
PHZYUQLIZKTSJE-UHFFFAOYSA-N | |
2755407 | |
OC(=O)COCCN1C(=O)C2=CC=CC=C2C1=O |
C12H11NO5 | |
MFCD06661364 | |
2-2-1,3-dioxoisoindolin-2-yl ethoxy acetic acid, 2-2-phthalimidoethoxy acetic acid, 2-2-phthalimidoethoxy-acetic acid, 2-1,3-dioxoisoindol-2-yl ethoxy acetic acid, 2-2-1,3-dioxoisoindol-2-yl ethoxy acetic acid, 2-phthalimido ethoxyacetic acid, 2-1,3-dioxoisoindolin-2-yl ethoxy acetic acid, 2-2-1,3-dioxoisoindolin-2-yl ethoxy aceticacid, 2-1,3-dioxo-1,3-dihydro-2h-isoindol-2-yl ethoxy acetic acid, 2-2-1,3-dioxo-2,3-dihydro-1h-isoindol-2-yl ethoxy acetic acid | |
2-[2-(1,3-dioxoisoindol-2-yl)ethoxy]acetic acid |
Specifications
69676-65-9 | |
C12H11NO5 | |
1 g | |
PHZYUQLIZKTSJE-UHFFFAOYSA-N | |
2-[2-(1,3-dioxoisoindol-2-yl)ethoxy]acetic acid | |
2755407 | |
97% |
130°C to 132°C | |
MFCD06661364 | |
2-2-1,3-dioxoisoindolin-2-yl ethoxy acetic acid, 2-2-phthalimidoethoxy acetic acid, 2-2-phthalimidoethoxy-acetic acid, 2-1,3-dioxoisoindol-2-yl ethoxy acetic acid, 2-2-1,3-dioxoisoindol-2-yl ethoxy acetic acid, 2-phthalimido ethoxyacetic acid, 2-1,3-dioxoisoindolin-2-yl ethoxy acetic acid, 2-2-1,3-dioxoisoindolin-2-yl ethoxy aceticacid, 2-1,3-dioxo-1,3-dihydro-2h-isoindol-2-yl ethoxy acetic acid, 2-2-1,3-dioxo-2,3-dihydro-1h-isoindol-2-yl ethoxy acetic acid | |
OC(=O)COCCN1C(=O)C2=CC=CC=C2C1=O | |
249.22 | |
249.22 | |
2-(2-Phthalimidoethoxy)acetic acid |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only