missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Hydroxyphenyl)benzothiazole 98.0+%, TCI America™

$69.92 - $277.79

Chemical Identifiers

CAS 3411-95-8
Molecular Formula C13H9NOS
Molecular Weight (g/mol) 227.28
MDL Number MFCD00022869
InChI Key NSDGEQWRYXOZLN-UKTHLTGXSA-N
Synonym phenol, o-2-benzothiazolyl, 2-2-hydroxyphenyl benzothiazole, o-2-benzothiazolyl phenol, phenol, 2-2-benzothiazolyl, 2-2-benzothiazolyl phenol, 2-o-hydroxyphenyl benzothiazole, 2-2'-hydroxyphenyl benzothiazole, 2-1,3-benzothiazol-2-yl phenol, 6z-6-3h-1,3-benzothiazol-2-ylidene cyclohexa-2,4-dien-1-one, mvvgspcxhrfddr-uhfffaoysa-n
PubChem CID 5376551
IUPAC Name 6-[(2E)-2,3-dihydro-1,3-benzothiazol-2-ylidene]cyclohexa-2,4-dien-1-one
SMILES O=C1C=CC=C\C1=C1\NC2=CC=CC=C2S1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
H09735G
View Documents
TCI America
H09735G
5 g
Each for $69.92
Only null left
 
H097325G
View Documents
TCI America
H097325G
25 g
Each for $277.79
Only null left
 
Description

Description

Specifications

Chemical Identifiers

3411-95-8
227.28
NSDGEQWRYXOZLN-UKTHLTGXSA-N
5376551
O=C1C=CC=C\C1=C1\NC2=CC=CC=C2S1
C13H9NOS
MFCD00022869
phenol, o-2-benzothiazolyl, 2-2-hydroxyphenyl benzothiazole, o-2-benzothiazolyl phenol, phenol, 2-2-benzothiazolyl, 2-2-benzothiazolyl phenol, 2-o-hydroxyphenyl benzothiazole, 2-2'-hydroxyphenyl benzothiazole, 2-1,3-benzothiazol-2-yl phenol, 6z-6-3h-1,3-benzothiazol-2-ylidene cyclohexa-2,4-dien-1-one, mvvgspcxhrfddr-uhfffaoysa-n
6-[(2E)-2,3-dihydro-1,3-benzothiazol-2-ylidene]cyclohexa-2,4-dien-1-one

Specifications

3411-95-8
C13H9NOS
5 g
NSDGEQWRYXOZLN-UKTHLTGXSA-N
6-[(2E)-2,3-dihydro-1,3-benzothiazol-2-ylidene]cyclohexa-2,4-dien-1-one
5376551
≥98.0% (HPLC,T)
2-(2-Hydroxyphenyl)benzothiazole
133°C
MFCD00022869
phenol, o-2-benzothiazolyl, 2-2-hydroxyphenyl benzothiazole, o-2-benzothiazolyl phenol, phenol, 2-2-benzothiazolyl, 2-2-benzothiazolyl phenol, 2-o-hydroxyphenyl benzothiazole, 2-2'-hydroxyphenyl benzothiazole, 2-1,3-benzothiazol-2-yl phenol, 6z-6-3h-1,3-benzothiazol-2-ylidene cyclohexa-2,4-dien-1-one, mvvgspcxhrfddr-uhfffaoysa-n
O=C1C=CC=C\C1=C1\NC2=CC=CC=C2S1
227.28
227.28
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

RTECSNumber : SJ7360000

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'