missing translation for 'onlineSavingsMsg'
Learn More

2,2-Dimethoxypropane, 98%

CAS: 77-76-9 | C5H12O2 | 104.149 g/mol

Supplier:  Thermo Scientific Chemicals A138100F

 View more versions of this product

Catalog No. AAA138100F


Only null left
Explore available promotions

Description

Description

2,2-Dimethoxypropane acts as a dehydrating agent. It also serves as an intermediate in the synthesis of vitamin E, vitamin A and various carotenoids such as astaxanthin. It is used as a reagent for the preparation of 1,2-diols, acetonides, isopropylidene derivatives of sugars, nucleosides, methyl esters of amino acids and enol ethers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2-Dimethoxypropane acts as a dehydrating agent. It also serves as an intermediate in the synthesis of vitamin E, vitamin A and various carotenoids such as astaxanthin. It is used as a reagent for the preparation of 1,2-diols, acetonides, isopropylidene derivatives of sugars, nucleosides, methyl esters of amino acids and enol ethers.

Solubility
Soluble in benzene, carbon tetrachloride, ethyl ether and n-butane, methanol. Moderately soluble in water.

Notes
Incompatible with oxidizing agents and acids.
Specifications

Specifications

2,2-Dimethoxypropane
-47°C
79°C to 81°C
98%
C5H12O2
MFCD00008479
UN3271
acetone dimethyl acetal, propane, 2,2-dimethoxy, acetone dimethyl ketal, acetone, dimethyl acetal, acetone-dimethyl acetal, 2,2-dimethoxy propane, acetone dimethylacetal, 2,2-dimethyoxypropane, 2,2-dimethoxy-propane, dimethoxypropan
HEWZVZIVELJPQZ-UHFFFAOYSA-N
2,2-dimethoxypropane
6495
98%
77-76-9
0.848
−11°C (12°F)
Sweet
1.378
2500 mL
635678
Soluble in benzene,carbon tetrachloride,ethyl ether and n-butane,methanol. Moderately soluble in water.
CC(C)(OC)OC
104.149
104.15
Liquid

Chemical Identifiers

77-76-9
104.149
HEWZVZIVELJPQZ-UHFFFAOYSA-N
6495
CC(C)(OC)OC
C5H12O2
MFCD00008479
acetone dimethyl acetal, propane, 2,2-dimethoxy, acetone dimethyl ketal, acetone, dimethyl acetal, acetone-dimethyl acetal, 2,2-dimethoxy propane, acetone dimethylacetal, 2,2-dimethyoxypropane, 2,2-dimethoxy-propane, dimethoxypropan
2,2-dimethoxypropane
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H319
Highly flammable liquid and vapour.
Causes serious eye irritation.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c

H225-H315-H319-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ETHERS, N.O.S.

EINECSNumber : 201-056-0

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only