missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,2'-Bipyridyl 99.0+%, TCI America™
$91.33 - $868.65
Chemical Identifiers
CAS | 366-18-7 |
---|---|
Molecular Formula | C10H8N2 |
Molecular Weight (g/mol) | 156.19 |
MDL Number | MFCD00006212 |
InChI Key | ROFVEXUMMXZLPA-UHFFFAOYSA-N |
Synonym | 2,2'-bipyridine, 2,2'-dipyridyl, 2,2'-bipyridyl, bipyridine, 2,2'-dipyridine, 2-2-pyridyl pyridine, 2,2'-bipyridin, alpha,alpha'-dipyridyl, alpha,alpha'-bipyridyl, 2,2-bipyridyl |
PubChem CID | 1474 |
ChEBI | CHEBI:30351 |
IUPAC Name | 2,2'-bipyridine |
SMILES | C1=CC=C(N=C1)C1=CC=CC=N1 |
Chemical Identifiers
366-18-7 | |
156.19 | |
ROFVEXUMMXZLPA-UHFFFAOYSA-N | |
1474 | |
2,2'-bipyridine |
C10H8N2 | |
MFCD00006212 | |
2,2'-bipyridine, 2,2'-dipyridyl, 2,2'-bipyridyl, bipyridine, 2,2'-dipyridine, 2-2-pyridyl pyridine, 2,2'-bipyridin, alpha,alpha'-dipyridyl, alpha,alpha'-bipyridyl, 2,2-bipyridyl | |
CHEBI:30351 | |
C1=CC=C(N=C1)C1=CC=CC=N1 |
Specifications
366-18-7 | |
White-Yellow | |
C10H8N2 | |
25 g | |
2,2'-bipyridine, 2,2'-dipyridyl, 2,2'-bipyridyl, bipyridine, 2,2'-dipyridine, 2-2-pyridyl pyridine, 2,2'-bipyridin, alpha,alpha'-dipyridyl, alpha,alpha'-bipyridyl, 2,2-bipyridyl | |
C1=CC=C(N=C1)C1=CC=CC=N1 | |
156.19 | |
CHEBI:30351 | |
≥99.0% (T) | |
2,2′-Bipyridyl |
72°C | |
273°C | |
MFCD00006212 | |
2811 | |
ROFVEXUMMXZLPA-UHFFFAOYSA-N | |
2,2'-bipyridine | |
1474 | |
156.19 | |
Crystalline Powder |
Safety and Handling
EINECSNumber : (5)-3723
RTECSNumber : DW1750000
TSCA : Yes