missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,2'-Bicinchoninic Acid Dipotassium Salt Hydrate 98.0+%, TCI America™
$76.43 - $223.68
Chemical Identifiers
CAS | 63451-34-3 |
---|---|
Molecular Formula | C20H12K2N2O4 |
Molecular Weight (g/mol) | 422.523 |
MDL Number | MFCD00014664 |
InChI Key | HRNSKWTYGVQLDN-UHFFFAOYSA-N |
PubChem CID | 66596657 |
IUPAC Name | 2-(4-carboxyquinolin-2-yl)quinoline-4-carboxylic acid;potassium |
SMILES | C1=CC=C2C(=C1)C(=CC(=N2)C3=NC4=CC=CC=C4C(=C3)C(=O)O)C(=O)O.[K].[K] |
Chemical Identifiers
63451-34-3 | |
422.523 | |
HRNSKWTYGVQLDN-UHFFFAOYSA-N | |
2-(4-carboxyquinolin-2-yl)quinoline-4-carboxylic acid;potassium |
C20H12K2N2O4 | |
MFCD00014664 | |
66596657 | |
C1=CC=C2C(=C1)C(=CC(=N2)C3=NC4=CC=CC=C4C(=C3)C(=O)O)C(=O)O.[K].[K] |
Specifications
63451-34-3 | |
MFCD00014664 | |
2,2′C-Biquinoline-4,4′C-dicarboxylic Acid Dipotassium Salt, 4,4′C-Dicarboxy-2,2′C-biquinoline Dipotassium Salt | |
C1=CC=C2C(=C1)C(=CC(=N2)C3=NC4=CC=CC=C4C(=C3)C(=O)O)C(=O)O.[K].[K] | |
422.523 | |
420.51 | |
Crystalline Powder |
C20H12K2N2O4 | |
1 g | |
HRNSKWTYGVQLDN-UHFFFAOYSA-N | |
2-(4-carboxyquinolin-2-yl)quinoline-4-carboxylic acid;potassium | |
66596657 | |
≥98.0% (HPLC) | |
2,2′-Bicinchoninic Acid Dipotassium Salt Hydrate [for Determination of Cu] |
Safety and Handling
TSCA : Yes