missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,2':5',2″-Terthiophene (purified by sublimation) 98.0+%, TCI America™
$132.35 - $395.17
Chemical Identifiers
CAS | 1081-34-1 |
---|---|
Molecular Formula | C12H8S3 |
Molecular Weight (g/mol) | 248.376 |
MDL Number | MFCD00012167 |
InChI Key | KXSFECAJUBPPFE-UHFFFAOYSA-N |
Synonym | 2,2':5',2-terthiophene, alpha-terthienyl, alpha-terthiophene, 2,5-di 2-thienyl thiophene, 2,2',5',2-terthienyl, 2,5-bis 2-thienyl thiophene, unii-0p77rau2rr, 2,2'-5',2-terthiophene, 0p77rau2rr, 2,2':5',2-terthienyl |
PubChem CID | 65067 |
ChEBI | CHEBI:10335 |
IUPAC Name | 2,5-dithiophen-2-ylthiophene |
SMILES | C1=CSC(=C1)C2=CC=C(S2)C3=CC=CS3 |
Chemical Identifiers
1081-34-1 | |
248.376 | |
KXSFECAJUBPPFE-UHFFFAOYSA-N | |
65067 | |
2,5-dithiophen-2-ylthiophene |
C12H8S3 | |
MFCD00012167 | |
2,2':5',2-terthiophene, alpha-terthienyl, alpha-terthiophene, 2,5-di 2-thienyl thiophene, 2,2',5',2-terthienyl, 2,5-bis 2-thienyl thiophene, unii-0p77rau2rr, 2,2'-5',2-terthiophene, 0p77rau2rr, 2,2':5',2-terthienyl | |
CHEBI:10335 | |
C1=CSC(=C1)C2=CC=C(S2)C3=CC=CS3 |
Specifications
1081-34-1 | |
C12H8S3 | |
1 g | |
KXSFECAJUBPPFE-UHFFFAOYSA-N | |
2,5-dithiophen-2-ylthiophene | |
65067 | |
248.38 | |
Crystalline Powder |
95°C | |
MFCD00012167 | |
2,2':5',2-terthiophene, alpha-terthienyl, alpha-terthiophene, 2,5-di 2-thienyl thiophene, 2,2',5',2-terthienyl, 2,5-bis 2-thienyl thiophene, unii-0p77rau2rr, 2,2'-5',2-terthiophene, 0p77rau2rr, 2,2':5',2-terthienyl | |
C1=CSC(=C1)C2=CC=C(S2)C3=CC=CS3 | |
248.376 | |
CHEBI:10335 | |
≥98.0% (GC) | |
2,2′:5′,2′′-Terthiophene (purified by sublimation) |
Safety and Handling
RTECSNumber : WZ9717750
TSCA : No