missing translation for 'onlineSavingsMsg'
Learn More

2,2,4-Trimethylpentane, ACS, 99%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor T11251LTGL

 View more versions of this product

Catalog No. 18608164


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

540-84-1
1 L
1e-05
ACS
1
Amber Glass Bottle
0.99

Chemical Identifiers

540-84-1
Product Suggestions

Product Suggestions

Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.