missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(1-Pyrazolyl)pyridine 98.0+%, TCI America™
$175.75 - $604.73
Chemical Identifiers
CAS | 25700-11-2 |
---|---|
Molecular Formula | C8H7N3 |
Molecular Weight (g/mol) | 145.17 |
MDL Number | MFCD09033534 |
InChI Key | XXTPHXNBKRVYJI-UHFFFAOYSA-N |
PubChem CID | 5324086 |
IUPAC Name | 2-(1H-pyrazol-1-yl)pyridine |
SMILES | C1=CN(N=C1)C1=CC=CC=N1 |
Chemical Identifiers
25700-11-2 | |
145.17 | |
XXTPHXNBKRVYJI-UHFFFAOYSA-N | |
2-(1H-pyrazol-1-yl)pyridine |
C8H7N3 | |
MFCD09033534 | |
5324086 | |
C1=CN(N=C1)C1=CC=CC=N1 |
Specifications
25700-11-2 | |
White | |
MFCD09033534 | |
XXTPHXNBKRVYJI-UHFFFAOYSA-N | |
2-(1H-pyrazol-1-yl)pyridine | |
5324086 | |
≥98.0% (GC) | |
2-(1-Pyrazolyl)pyridine |
39°C | |
C8H7N3 | |
1 g | |
C1=CN(N=C1)C1=CC=CC=N1 | |
145.17 | |
145.17 | |
Crystalline Lumps |
Safety and Handling
TSCA : No