missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(1S,3R)-(-)-Camphoric acid, 98%
CAS: 560-09-8 | C10H16O4 | 200.234 g/mol
$79.27 - $990.34
Chemical Identifiers
CAS | 560-09-8 |
---|---|
Molecular Formula | C10H16O4 |
Molecular Weight (g/mol) | 200.234 |
MDL Number | MFCD00064937 |
InChI Key | LSPHULWDVZXLIL-QUBYGPBYSA-N |
Synonym | 1s,3r-1,2,2-trimethylcyclopentane-1,3-dicarboxylic acid, #NAME?, camphoric acid, camphoric acid,-, 1s,3r-camphoric acid, 1s,3r---camphoric acid, 1s,3r-1,2,2-trimethyl-1,3-cyclopentanedicarboxylic acid, 1,3-cyclopentanedicarboxylic acid, 1,2,2-trimethyl-, 1s,3r, l-camphoric acid, d-+-camphoric acid |
PubChem CID | 219463 |
IUPAC Name | (1S,3R)-1,2,2-trimethylcyclopentane-1,3-dicarboxylic acid |
SMILES | CC1(C(CCC1(C)C(=O)O)C(=O)O)C |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH54042MD
|
Thermo Scientific Chemicals
H54042MD |
250 mg |
Each for $79.27
|
|
|||||
AAH5404203
|
Thermo Scientific Chemicals
H5404203 |
1 g |
Each for $204.78
|
|
|||||
AAH5404206
|
Thermo Scientific Chemicals
H5404206 |
5 g |
Each for $990.34
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
560-09-8 | |
200.234 | |
LSPHULWDVZXLIL-QUBYGPBYSA-N | |
219463 | |
CC1(C(CCC1(C)C(=O)O)C(=O)O)C |
C10H16O4 | |
MFCD00064937 | |
1s,3r-1,2,2-trimethylcyclopentane-1,3-dicarboxylic acid, #NAME?, camphoric acid, camphoric acid,-, 1s,3r-camphoric acid, 1s,3r---camphoric acid, 1s,3r-1,2,2-trimethyl-1,3-cyclopentanedicarboxylic acid, 1,3-cyclopentanedicarboxylic acid, 1,2,2-trimethyl-, 1s,3r, l-camphoric acid, d-+-camphoric acid | |
(1S,3R)-1,2,2-trimethylcyclopentane-1,3-dicarboxylic acid |
Specifications
188°C to 190°C | |
C10H16O4 | |
1874674 | |
1s,3r-1,2,2-trimethylcyclopentane-1,3-dicarboxylic acid, #NAME?, camphoric acid, camphoric acid,-, 1s,3r-camphoric acid, 1s,3r---camphoric acid, 1s,3r-1,2,2-trimethyl-1,3-cyclopentanedicarboxylic acid, 1,3-cyclopentanedicarboxylic acid, 1,2,2-trimethyl-, 1s,3r, l-camphoric acid, d-+-camphoric acid | |
CC1(C(CCC1(C)C(=O)O)C(=O)O)C | |
200.234 | |
200.23 | |
250 mg | |
(1S,3R)-(-)-Camphoric acid |
560-09-8 | |
MFCD00064937 | |
141733 | |
LSPHULWDVZXLIL-QUBYGPBYSA-N | |
(1S,3R)-1,2,2-trimethylcyclopentane-1,3-dicarboxylic acid | |
219463 | |
98% | |
-48° (c=10 in Ethanol) |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only