missing translation for 'onlineSavingsMsg'
Learn More
Learn More
National Safety Apparel™ 12 Cal UltraSoft FR Coveralls 28 in. Inseam
For enhanced protection Westex™ UltraSoft is the premium choice for superior comfort, improved safety and above average durability. With a unique blend of 88% cotton and 12% high-tenacity nylon UltraSoft garments provide a smooth feel for increased comfort and the inherently FR material delivers exceptional performance in reliability
Supplier: National Safety Apparel™ C88UP2X28
Description
- Arc Rating: 12 cal/cm²
- CAT 2
- Fabric/Material: 9 oz. Westex™ UltraSoft™
- Comfortable sewn-in elastic waist for better fit
- Dual certified to protect against flash fire and arc flash hazards
- 2-way covered zipper front for easy entry
- Two front chest pockets with flaps and hidden snap closures
- Large reinforced back pockets
- Convenient 6 pocket style with side pass through access
- Leg openings made to fit over work boots
- All metal covered to ensure no contact with skin
- Action back allows for full range of motion
- FR fabric will not melt, drip, or ignite
- Zip front closure with additional covered snaps
- Home laundry
- Made in the USA
Specifications
2X-Large | |
Navy | |
Zipper | |
Arc Rating: 12 cal/cm², CAT 2, ASTM F1506, OSHA 1910.269, NFPA 70 E, NFPA 2112, CSA Z462 |
Cotton, Nylon | |
Open | |
Stitched | |
None |