missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ 10% Neutral Buffered Formalin in Seal-Rite Containers

Use these containers for the transportation and short-term storage of patient specimens.

Supplier:  Epredia™ 51201

 View more versions of this product

Catalog No. 36-101-5672


Only null left
Explore available promotions

Description

Description

Features:
  • Seal-Rite containers are engineered to maintain seal at 95KPA – the industry standard for proper sealing and transportation via air shipment
  • Double-click feature provides audible confirmation that the lid has been aligned, tightened properly and sealed
  • Arrow and lock icons align to visually signify the containers have been properly sealed
  • Embossed mL measuring scale identifies volume of liquid in the container
  • Knurled cap and base of the jar for better gripping
  • Safety. Security. Peace of Mind – Avoid the loss of precious tissue samples and exposure of staff to chemicals caused by leaking containers
A properly closed Epredia Seal-Rite container is leak-proof.
The containers have been tested using industry-standard International Safe Transit Association (ISTA) performance validation. Internal tests were performed as well with 0% leakage rate.
Specifications

Specifications

20 mL
Carton
Formalin
192/cs
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions