missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epredia™ 10% Neutral Buffered Formalin in Seal-Rite Containers
Use these containers for the transportation and short-term storage of patient specimens.
Supplier: Epredia™ 9990910
Description
- Phosphate-free
- Use for storage of all tissue types
- Stores specimens without excessive shrinking or hardening
- Available premixed, ready-to-use, or in economical concentrate
- Concentrate is not affected by freezing, reduces shelf space, and is less expensive to ship
- Pre-filled, leak-resistant cups are available
Specifications
20 mL | |
Tray | |
Specimen Storage Containers | |
180/cs |