missing translation for 'onlineSavingsMsg'
Learn More
Learn More
10,10'-Dibromo-9,9'-bianthracene 98.0+%, TCI America™
$246.05 - $740.03
Chemical Identifiers
CAS | 121848-75-7 |
---|---|
Molecular Formula | C28H16Br2 |
Molecular Weight (g/mol) | 512.244 |
MDL Number | MFCD11045035 |
InChI Key | NPNNLGXEAGTSRN-UHFFFAOYSA-N |
PubChem CID | 15195999 |
IUPAC Name | 9-bromo-10-(10-bromoanthracen-9-yl)anthracene |
SMILES | C1=CC=C2C(=C1)C(=C3C=CC=CC3=C2Br)C4=C5C=CC=CC5=C(C6=CC=CC=C64)Br |
Chemical Identifiers
121848-75-7 | |
512.244 | |
NPNNLGXEAGTSRN-UHFFFAOYSA-N | |
9-bromo-10-(10-bromoanthracen-9-yl)anthracene |
C28H16Br2 | |
MFCD11045035 | |
15195999 | |
C1=CC=C2C(=C1)C(=C3C=CC=CC3=C2Br)C4=C5C=CC=CC5=C(C6=CC=CC=C64)Br |
Specifications
121848-75-7 | |
Yellow | |
MFCD11045035 | |
NPNNLGXEAGTSRN-UHFFFAOYSA-N | |
9-bromo-10-(10-bromoanthracen-9-yl)anthracene | |
15195999 | |
≥98.0% (HPLC,T) | |
10,10′-Dibromo-9,9′-bianthracene |
342°C | |
C28H16Br2 | |
1 g | |
C1=CC=C2C(=C1)C(=C3C=CC=CC3=C2Br)C4=C5C=CC=CC5=C(C6=CC=CC=C64)Br | |
512.244 | |
512.24 | |
Crystalline Powder |
Safety and Handling
TSCA : No