missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Propanol, Analytical Standard, ≥99.9% (GC), MilliporeSigma™ Supelco™
Supplier: Merck Emd Millipore 9656610MLF
Specifications
71-23-8 | |
2.1 (vs. air) | |
Neat | |
MFCD00002941 | |
97°C | |
10 mL | |
≥99.9% (GC) |
10 mmHg (147°C) | |
0.804 g/mL (at 25°C) | |
CH3CH2CH2OH | |
-127°C | |
Propyl alcohol | |
60.10 | |
Analytical Standard |