missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Octanol, ACS reagent, ≥99%, Honeywell™ Riedel-de Haën™
ACS Reagent,≥99%
$126.80 - $231.58
Chemical Identifiers
CAS | 111-87-5 |
---|---|
Molecular Formula | C8H18O |
Molecular Weight (g/mol) | 130.23 |
MDL Number | MFCD00002988 |
InChI Key | KBPLFHHGFOOTCA-UHFFFAOYSA-N |
Synonym | 1-octanol, octanol, octyl alcohol, n-octanol, capryl alcohol, caprylic alcohol, n-octyl alcohol, heptyl carbinol, 1-hydroxyoctane, primary octyl alcohol |
PubChem CID | 957 |
ChEBI | CHEBI:16188 |
IUPAC Name | octan-1-ol |
SMILES | CCCCCCCCO |
Description
ACS GENERAL USE SOLVENTS
- value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
- suitable for less demanding applications such as routine isocratic HPLC
- variations and decrease issues traceable to solvent contamination
- all solvents are tested to meet ACS general use specifications
Chemical Identifiers
111-87-5 | |
130.23 | |
KBPLFHHGFOOTCA-UHFFFAOYSA-N | |
957 | |
octan-1-ol |
C8H18O | |
MFCD00002988 | |
1-octanol, octanol, octyl alcohol, n-octanol, capryl alcohol, caprylic alcohol, n-octyl alcohol, heptyl carbinol, 1-hydroxyoctane, primary octyl alcohol | |
CHEBI:16188 | |
CCCCCCCCO |
Specifications
111-87-5 | |
0.827g/mL at 25°C | |
80°C (176°F) | |
Glass Bottle | |
n20/D 1.429 | |
MFCD00002988 | |
1697461 | |
KBPLFHHGFOOTCA-UHFFFAOYSA-N | |
octan-1-ol | |
4.5 (vs air) | |
CHEBI:16188 | |
≥99% | |
Liquid |
-15°C | |
194°C | |
Characteristic | |
C8H18O | |
CH3(CH2)7OH | |
100 mL | |
1-octanol, octanol, octyl alcohol, n-octanol, capryl alcohol, caprylic alcohol, n-octyl alcohol, heptyl carbinol, 1-hydroxyoctane, primary octyl alcohol | |
CCCCCCCCO | |
130.23 | |
957 | |
130.23g/mol | |
ACS Reagent | |
1-Octanol |
Safety and Handling
P273-P280-P305 + P351 + P338-P337 + P313
RTECSNumber : 203-917-6