missing translation for 'onlineSavingsMsg'
Learn More

1-Octanesulfonic Acid Sodium Salt, Monohydrate, Reagent HPLC, 99%, Spectrum™ Chemical

O1001, 207596-29-0, C8H17NaO3S·H2O

$267.83 - $23490.46

Chemical Identifiers

CAS 207596-29-0
Molecular Formula C8H19NaO4S
Molecular Weight (g/mol) 234.29
MDL Number MFCD00149551
InChI Key MBURIAHQXJQKRE-UHFFFAOYSA-M
IUPAC Name sodium hydrate octane-1-sulfonate
SMILES O.[Na+].CCCCCCCCS([O-])(=O)=O
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18600068
View Documents
Spectrum Chemical Mfg Cor
O10015GM
5 g Amber Glass Bottle
Each for $267.83
Only null left
 
18600069
View Documents
Spectrum Chemical Mfg Cor
O100125GM
25 g Amber Glass Bottle
Each for $855.77
Only null left
 
18600070
View Documents
Spectrum Chemical Mfg Cor
O1001100GM
100 g Amber Glass Bottle
Each for $2,463.29
Only null left
 
18600071
View Documents
Spectrum Chemical Mfg Cor
O10015KGBL
5 kg Fiber Drum N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Spectrum™ Chemical 1-Octanesulfonic Acid Sodium Salt, Monohydrate, Reagent HPLC is an ion pairing reagent which will enhance the retention of polar, ionizable compounds such as peptides and proteins. The Reagent grade denotes that this chemical is the highest quality commercially available and that the American Chemical Society has not officially set any specifications for this material. Spectrum Chemical manufactured Reagent grade products meet the toughest regulatory standards for quality and purity.
Specifications

Chemical Identifiers

207596-29-0
234.29
MBURIAHQXJQKRE-UHFFFAOYSA-M
O.[Na+].CCCCCCCCS([O-])(=O)=O
C8H19NaO4S
MFCD00149551
sodium hydrate octane-1-sulfonate

Specifications

207596-29-0
Amber Glass Bottle
MFCD00149551
MBURIAHQXJQKRE-UHFFFAOYSA-M
sodium hydrate octane-1-sulfonate
99%
100%
C8H19NaO4S
5 g
O.[Na+].CCCCCCCCS([O-])(=O)=O
234.29
Reagent
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'