missing translation for 'onlineSavingsMsg'
Learn More

1-Naphthol, 99%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor N100925KGBL

 View more versions of this product

Catalog No. 18608128

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

Specifications

Specifications

90-15-3
288°C
Amber Glass Bottle
0.99
Crystalline Powder or Lumps
1
92° to 99°C
25 kg
Reagent

Chemical Identifiers

90-15-3
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.