Learn More
Eisco™ 1 Hole ASTM Neoprene Stoppers
Pack of ten 1-hole ASTM neoprene stoppers. Made from long lasting neoprene rubber. Oil resistant, use up to 250°F / 120°C. Suitable for use with petroleum products, minerals, oils, fats as well as most inorganic acids.
$11.13 - $1542.67
Specifications
Certifications/Compliance | ASTM |
---|---|
Autoclavable | Autoclavable |
Color | Blue |
Material | Neoprene |
Product Type | Steri-stopper |
Catalog Number | Mfr. No. | Diameter (Metric) Bottom | Diameter (Metric) | Diameter (Metric) Hole | Height (Metric) | Price | Quantity | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Diameter (Metric) Bottom | Diameter (Metric) | Diameter (Metric) Hole | Height (Metric) | Price | Quantity | ||
S41056
|
Eisco™
CH0321A1H |
8.2 mm | 12.7 mm | 4 mm | 25 mm |
Pack of 10 Each for $11.13
|
|
||
S27039
|
Eisco™
CH0321N1H |
36 mm | 41 mm | 5 mm | 21 mm |
Each for $113.33
|
|
||
S41058
|
Eisco™
CH0321B1H |
10 mm | 15 mm | 4 mm | 25 mm |
Pack of 10 Each for $14.60
|
|
||
S41060
|
Eisco™
CH0321C1H |
13 mm | 17 mm | 4 mm | 25 mm |
Pack of 10 Each for $21.07
|
|
||
S41063
|
Eisco™
CH0321D1H |
14 mm | 19 mm | 4 mm | 25 mm |
Pack of 10 Each for $23.10
|
|
||
S41067
|
Eisco™
CH0321E1H |
16 mm | 20 mm | 5 mm | 25 mm |
Pack of 10 Each for $21.90
|
|
||
S41070
|
Eisco™
CH0321F1H |
18 mm | 24 mm | 5 mm | 25 mm |
Pack of 10 Each for $28.37
|
|
||
S41073
|
Eisco™
CH0321G1H |
20 mm | 26 mm | 5 mm | 25 mm |
Pack of 10 Each for $32.30
|
|
||
S41076
|
Eisco™
CH0321H1H |
23 mm | 27 mm | 5 mm | 25 mm |
Pack of 10 Each for $38.67
|
|
||
S41079
|
Eisco™
CH0321I1H |
24 mm | 28 mm | 5 mm | 25 mm |
Pack of 10 Each for $51.63
|
|
||
S41082
|
Eisco™
CH0321J1H |
26 mm | 32 mm | 5 mm | 25 mm |
Pack of 10 Each for $97.13
|
|
||
S41085
|
Eisco™
CH0321K1H |
27 mm | 34 mm | 5 mm | 25 mm |
Pack of 10 Each for $67.97
|
|
||
S41088
|
Eisco™
CH0321L1H |
30 mm | 37 mm | 5 mm | 25 mm |
Pack of 10 Each for $79.43
|
|
||
S41091
|
Eisco™
CH0321M1H |
31 mm | 39 mm | 5 mm | 25 mm |
Pack of 10 Each for $93.57
|
|
||
S41096
|
Eisco™
CH0321O1H |
36 mm | 43 mm | 5 mm | 25 mm |
Pack of 10 Each for $108.97
|
|
||
S41099
|
Eisco™
CH0321P1H |
37 mm | 45 mm | 5 mm | 25 mm |
Pack of 10 Each for $120.80
|
|
||
S41101
|
Eisco™
CH0321Q1H |
42 mm | 50 mm | 5 mm | 25 mm |
Pack of 10 Each for $158.33
|
|
||
S41104
|
Eisco™
CH0321R1H |
45 mm | 53 mm | 5 mm | 25 mm |
Pack of 10 Each for $162.30
|
|
||
S41107
|
Eisco™
CH0321S1H |
48 mm | 56 mm | 5 mm | 25 mm |
Pack of 10 Each for $189.87
|
|
||
S41110
|
Eisco™
CH0321T1H |
50 mm | 63 mm | 5 mm | 25 mm |
Pack of 10 Each for $223.53
|
|
||
S41113
|
Eisco™
CH0321U1H |
54 mm | 64 mm | 5 mm | 25 mm |
Pack of 10 Each for $241.17
|
|
||
S41117
|
Eisco™
CH0321V1H |
58 mm | 68 mm | 5 mm | 25 mm |
Pack of 10 Each for $271.20
|
|
||
S41120
|
Eisco™
CH0321W1H |
62 mm | 75 mm | 5 mm | 35 mm |
Pack of 10 Each for $470.03
|
|
||
S41125
|
Eisco™
CH0321X1H |
75 mm | 90 mm | 5 mm | 39 mm |
Pack of 10 Each for $614.67
|
|
||
S41128
|
Eisco™
CH0321Y1H |
83 mm | 103 mm | 5 mm | 39 mm |
Pack of 10 Each for $964.17
|
|
||
S41131
|
Eisco™
CH0321Z1H |
90 mm | 127 mm | 5 mm | 50 mm |
Pack of 10 Each for $1,542.67
|
|
||
Description
Warranty
1 year limited manufacturer warranty
Specifications
ASTM | |
Blue | |
Steri-stopper | |
Non-sterile | |
Stopper for Labware | |
-40 to 130°C |
Autoclavable | |
Neoprene | |
Conical | |
Reusable | |
Petroleum products, minerals, oils and fats, as well as most inorganic acids and bases |