missing translation for 'onlineSavingsMsg'
Learn More

1-Hexyl isothiocyanate, 97%

CAS: 4404-45-9 | C7H13NS | 143.248 g/mol

$104.67 - $363.66

Chemical Identifiers

CAS 4404-45-9
Molecular Formula C7H13NS
Molecular Weight (g/mol) 143.248
MDL Number MFCD00014445
InChI Key WXYAXKKXIGHXDS-UHFFFAOYSA-N
Synonym hexyl isothiocyanate, n-hexyl isothiocyanate, hexane, 1-isothiocyanato, 1-hexyl isothiocyanate, hexylisothiocyanate, isothiocyanic acid, hexyl ester, unii-2s7c3ga0z5, 1-hexylisothiocyanate, 1-isothiocyanato-hexane, hexanisothiocyanate
PubChem CID 78120
IUPAC Name 1-isothiocyanatohexane
SMILES CCCCCCN=C=S
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1222306
View Documents
Thermo Scientific Chemicals
A1222306
5 g
Each for $104.67
Only null left
 
AAA1222314
View Documents
Thermo Scientific Chemicals
A1222314
25 g
Each for $363.66
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

4404-45-9
143.248
WXYAXKKXIGHXDS-UHFFFAOYSA-N
78120
CCCCCCN=C=S
C7H13NS
MFCD00014445
hexyl isothiocyanate, n-hexyl isothiocyanate, hexane, 1-isothiocyanato, 1-hexyl isothiocyanate, hexylisothiocyanate, isothiocyanic acid, hexyl ester, unii-2s7c3ga0z5, 1-hexylisothiocyanate, 1-isothiocyanato-hexane, hexanisothiocyanate
1-isothiocyanatohexane

Specifications

4404-45-9
216°C
Pungent
1.4915
5 g
1751837
hexyl isothiocyanate, n-hexyl isothiocyanate, hexane, 1-isothiocyanato, 1-hexyl isothiocyanate, hexylisothiocyanate, isothiocyanic acid, hexyl ester, unii-2s7c3ga0z5, 1-hexylisothiocyanate, 1-isothiocyanato-hexane, hexanisothiocyanate
CCCCCCN=C=S
143.248
143.25
1-Hexyl isothiocyanate
0.933
90°C (194°F)
C7H13NS
MFCD00014445
UN2922
Moisture sensitive
WXYAXKKXIGHXDS-UHFFFAOYSA-N
1-isothiocyanatohexane
78120
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H331-H314-H318-H227-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Harmful if swallowed.
Harmful in contact with skin.

P210-P235-P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P311-P312-P330-P332+P313-P361-P363-P370+P378q-P501c

H227-H301+H311+H331-H315-H319-H335

DOTInformation : Hazard Class: 6.1; Packaging Group: III

EINECSNumber : 224-549-2

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only