missing translation for 'onlineSavingsMsg'
Learn More

1-Hexanesulfonic Acid Sodium Salt, Monohydrate, 98%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor H1032100GM

 View more versions of this product

Catalog No. 18608115


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

207300-91-2
Amber Glass Bottle
0.98
1
100 g
Reagent

Chemical Identifiers

207300-91-2
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.