missing translation for 'onlineSavingsMsg'
Learn More

1-Cyclohexyl-3-(2-morpholinoethyl)carbodiimide Metho-p-toluenesulfonate 96.0+%, TCI America™

$622.59 - $2486.65

Chemical Identifiers

CAS 2491-17-0
Molecular Formula C21H33N3O4S
Molecular Weight (g/mol) 423.57
MDL Number MFCD00011979
InChI Key GBCAVSYHPPARHX-UHFFFAOYSA-M
Synonym cme-carbodiimide, morpho cdi, 1-cyclohexyl 3-2-morpholinoethyl carbodiimide methotosylate, 4-2-cyclohexylimino methylene amino ethyl-4-methylmorpholin-4-ium 4-methylbenzenesulfonate, cmc, 1-cyclohexyl-3-2-morpholinoethyl carbodiimide metho-p-toluenesulfonate, n-cyclohexyl-n'-2-morpholinoethyl carbodiimide metho-p-toluenesulfonate, cmc n-cyclohexyl-n-2-morpholinoethyl carbodiimide metho-p-toluenesulfonate, chm van, 4-5-cyclohexyl-3,5-diazapenta-3,4-dienyl-4-methylmorpholine, 4-methylbenzene sulfonic acid
PubChem CID 17220
IUPAC Name 4-(2-{[(cyclohexylimino)methylidene]amino}ethyl)-4-methylmorpholin-4-ium 4-methylbenzene-1-sulfonate
SMILES CC1=CC=C(C=C1)S([O-])(=O)=O.C[N+]1(CCN=C=NC2CCCCC2)CCOCC1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
C07935G
View Documents
TCI America
C07935G
5 g
Each for $622.59
Only null left
 
C079325G
View Documents
TCI America
C079325G
25 g
Each for $2,486.65
Only null left
 
Description

Description

Specifications

Chemical Identifiers

2491-17-0
423.57
GBCAVSYHPPARHX-UHFFFAOYSA-M
17220
CC1=CC=C(C=C1)S([O-])(=O)=O.C[N+]1(CCN=C=NC2CCCCC2)CCOCC1
C21H33N3O4S
MFCD00011979
cme-carbodiimide, morpho cdi, 1-cyclohexyl 3-2-morpholinoethyl carbodiimide methotosylate, 4-2-cyclohexylimino methylene amino ethyl-4-methylmorpholin-4-ium 4-methylbenzenesulfonate, cmc, 1-cyclohexyl-3-2-morpholinoethyl carbodiimide metho-p-toluenesulfonate, n-cyclohexyl-n'-2-morpholinoethyl carbodiimide metho-p-toluenesulfonate, cmc n-cyclohexyl-n-2-morpholinoethyl carbodiimide metho-p-toluenesulfonate, chm van, 4-5-cyclohexyl-3,5-diazapenta-3,4-dienyl-4-methylmorpholine, 4-methylbenzene sulfonic acid
4-(2-{[(cyclohexylimino)methylidene]amino}ethyl)-4-methylmorpholin-4-ium 4-methylbenzene-1-sulfonate

Specifications

2491-17-0
C21H33N3O4S
5 g
GBCAVSYHPPARHX-UHFFFAOYSA-M
4-(2-{[(cyclohexylimino)methylidene]amino}ethyl)-4-methylmorpholin-4-ium 4-methylbenzene-1-sulfonate
17220
≥96.0% (N,T)
1-Cyclohexyl-3-(2-morpholinoethyl)carbodiimide Metho-p-toluenesulfonate [for Peptide Synthesis]
White-Yellow
MFCD00011979
cme-carbodiimide, morpho cdi, 1-cyclohexyl 3-2-morpholinoethyl carbodiimide methotosylate, 4-2-cyclohexylimino methylene amino ethyl-4-methylmorpholin-4-ium 4-methylbenzenesulfonate, cmc, 1-cyclohexyl-3-2-morpholinoethyl carbodiimide metho-p-toluenesulfonate, n-cyclohexyl-n'-2-morpholinoethyl carbodiimide metho-p-toluenesulfonate, cmc n-cyclohexyl-n-2-morpholinoethyl carbodiimide metho-p-toluenesulfonate, chm van, 4-5-cyclohexyl-3,5-diazapenta-3,4-dienyl-4-methylmorpholine, 4-methylbenzene sulfonic acid
CC1=CC=C(C=C1)S([O-])(=O)=O.C[N+]1(CCN=C=NC2CCCCC2)CCOCC1
423.57
423.57
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'