missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Chlorobutane CHROMASOLV™, for HPLC, ≥99.8%, Honeywell Riedel-de Haën™
CHROMASOLV™, for HPLC,≥99.8%
$553.40 - $553.40
Chemical Identifiers
CAS | 109-69-3 |
---|---|
Molecular Formula | C4H9Cl |
Molecular Weight (g/mol) | 92.57 |
MDL Number | MFCD00001009 |
InChI Key | VFWCMGCRMGJXDK-UHFFFAOYSA-N |
Synonym | butyl chloride, n-butyl chloride, butane, 1-chloro, chlorure de butyle, nbc wormer, n-propylcarbinyl chloride, sure shot, butane, chloro, chlorobutane, n-chlorobutane |
PubChem CID | 8005 |
IUPAC Name | 1-chlorobutane |
SMILES | CCCCCl |
Description
- For HPLC and organic synthesis
- High-purity, multipurpose solvents
- High lot-to-lot consistency
Chemical Identifiers
109-69-3 | |
92.57 | |
VFWCMGCRMGJXDK-UHFFFAOYSA-N | |
8005 | |
CCCCCl |
C4H9Cl | |
MFCD00001009 | |
butyl chloride, n-butyl chloride, butane, 1-chloro, chlorure de butyle, nbc wormer, n-propylcarbinyl chloride, sure shot, butane, chloro, chlorobutane, n-chlorobutane | |
1-chlorobutane |
Specifications
109-69-3 | |
0.886g/mL at 25°C (lit.) | |
C4H9Cl | |
CH3(CH2)3Cl | |
2 L | |
butyl chloride, n-butyl chloride, butane, 1-chloro, chlorure de butyle, nbc wormer, n-propylcarbinyl chloride, sure shot, butane, chloro, chlorobutane, n-chlorobutane | |
CCCCCl | |
92.57 | |
92.57g/mol | |
≥99.8% | |
1-Chlorobutane, for HPLC |
-123°C | |
77°C to 78°C | |
n20/D 1.402 | |
MFCD00001009 | |
1730909 | |
VFWCMGCRMGJXDK-UHFFFAOYSA-N | |
1-chlorobutane | |
8005 | |
80.1mmHg (78.4°C) | |
CHROMASOLV™ |
Safety and Handling
P210-P273-P301 + P310-P331-P403 + P235
H225-H304-H336-H412
EINECSNumber : 203-696-6
RTECSNumber : EJ6300000