missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Butanol, Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur., ≥99.5% (GC), Honeywell Riedel-de Haën™
Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur.,≥99.5% (GC)
Supplier: Honeywell Chemicals 3306525L
Description
ACS GENERAL USE SOLVENTS
- value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
- suitable for less demanding applications such as routine isocratic HPLC
- variations and decrease issues traceable to solvent contamination
- all solvents are tested to meet ACS general use specifications
Specifications
1-Butanol | |
71-36-3 | |
0.81g/mL at 25°C | |
0.112 | |
649°F | |
n20/D 1.399 | |
CH3(CH2)3OH | |
UN1120 | |
1-butanol, butanol, n-butanol, butyl alcohol, n-butyl alcohol, 1-hydroxybutane, propylcarbinol, butyl hydroxide, propylmethanol, methylolpropane | |
LRHPLDYGYMQRHN-UHFFFAOYSA-N | |
butan-1-ol | |
263 | |
74.12g/mol | |
ACS/puriss. p.a. |
Reag. Ph. Eur. reag. ISO | |
Colorless | |
2.5 L | |
35°C (95°F) | |
Poly Bottle | |
C4H10O | |
MFCD00002964 | |
969148 | |
66g/L in water at 20°C, pH 7 | |
CCCCO | |
74.123 | |
CHEBI:28885 | |
≥99.5% (GC) |
Chemical Identifiers
71-36-3 | |
74.123 | |
LRHPLDYGYMQRHN-UHFFFAOYSA-N | |
263 | |
butan-1-ol |
C4H10O | |
MFCD00002964 | |
1-butanol, butanol, n-butanol, butyl alcohol, n-butyl alcohol, 1-hydroxybutane, propylcarbinol, butyl hydroxide, propylmethanol, methylolpropane | |
CHEBI:28885 | |
CCCCO |
Safety and Handling
P210-P280-P301 + P312 + P330-P304 + P340 + P312-P305 + P351 + P338 + P310-P403 + P235
H226-H302-H315-H318-H335-H336
Danger
DOTInformation : Class 3 - PG 3 - Butanols
EINECSNumber : 200-751-6
RTECSNumber : EO1400000