missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Bromopentadecane, 97%
CAS: 629-72-1 | C15H31Br | 291.32 g/mol
$146.00 - $487.22
Chemical Identifiers
CAS | 629-72-1 |
---|---|
Molecular Formula | C15H31Br |
Molecular Weight (g/mol) | 291.32 |
MDL Number | MFCD00000229 |
InChI Key | JKOTZBXSNOGCIF-UHFFFAOYSA-N |
Synonym | pentadecyl bromide, pentadecane, 1-bromo, 1-bromo pentadecane, n-pentadecyl bromide, pentadecane,1-bromo, 1-bromopentadecane, acmc-209nb8, dsstox_cid_29296, dsstox_rid_83414, dsstox_gsid_49339 |
PubChem CID | 12394 |
IUPAC Name | 1-bromopentadecane |
SMILES | CCCCCCCCCCCCCCCBr |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL0761809
|
Thermo Scientific Chemicals
L0761809 |
10 g |
Each for $146.00
|
|
|||||
AAL0761818
|
Thermo Scientific Chemicals
L0761818 |
50 g |
Each for $487.22
|
|
|||||
Description
1-Bromopentadecane has been used as regent in the synthesis of (S)-eicos-(4E)-en-1-yn-3-ol, a bioactive component of the marine sponge Cribrochalina vasculum.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-Bromopentadecane has been used as regent in the synthesis of (S)-eicos-(4E)-en-1-yn-3-ol, a bioactive component of the marine sponge Cribrochalina vasculum.
Solubility
Not miscible in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents. Incompatible materials are oxidizing agents.
Chemical Identifiers
629-72-1 | |
291.32 | |
JKOTZBXSNOGCIF-UHFFFAOYSA-N | |
12394 | |
CCCCCCCCCCCCCCCBr |
C15H31Br | |
MFCD00000229 | |
pentadecyl bromide, pentadecane, 1-bromo, 1-bromo pentadecane, n-pentadecyl bromide, pentadecane,1-bromo, 1-bromopentadecane, acmc-209nb8, dsstox_cid_29296, dsstox_rid_83414, dsstox_gsid_49339 | |
1-bromopentadecane |
Specifications
629-72-1 | |
1.005 | |
>110°C (230°F) | |
1.4608 | |
10 g | |
pentadecyl bromide, pentadecane, 1-bromo, 1-bromo pentadecane, n-pentadecyl bromide, pentadecane,1-bromo, 1-bromopentadecane, acmc-209nb8, dsstox_cid_29296, dsstox_rid_83414, dsstox_gsid_49339 | |
JKOTZBXSNOGCIF-UHFFFAOYSA-N | |
1-bromopentadecane | |
12394 | |
97% |
18°C to 19°C | |
159°C to 160°C (5 mmHg) | |
C15H31Br | |
MFCD00000229 | |
1744553 | |
Not miscible in water. | |
CCCCCCCCCCCCCCCBr | |
291.32 | |
291.32 | |
1-Bromopentadecane |
Safety and Handling
EINECSNumber : 211-104-2
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only