missing translation for 'onlineSavingsMsg'
Learn More

1-Bromobutane, 98%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor B11521KG

 View more versions of this product

Catalog No. 18608078


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

109-65-9
Amber Glass Bottle
0.98
1
1 kg
Reagent

Chemical Identifiers

109-65-9
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.