missing translation for 'onlineSavingsMsg'
Learn More

1,6-Hexanediamine, 98%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor HE17010KGBL

 View more versions of this product

Catalog No. 18608071

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

Specifications

Specifications

124-09-4
Steel Pail
0.98
1
10 kg
Reagent

Chemical Identifiers

124-09-4
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.