missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,5-Diphenylcarbazide, ≥98% (HPLC), Reag. Ph. Eur., MilliporeSigma™ Supelco™
1,5-Diphenylcarbazide reaction with chromium(VI), has been widely used for the spectro- photometric determination of chromium. It is also an electron donor, and can be used for measuring photosystem 2 activity in subchloroplast fragments.
$110.30 - $667.54
Chemical Identifiers
MDL Number | MFCD00003013 |
---|---|
Synonym | 1,5-Diphenylcarbohydrazide; sym.-Diphenylcarbazide |
Chemical Identifiers
MFCD00003013 | |
1,5-Diphenylcarbohydrazide; sym.-Diphenylcarbazide |
Specifications
170°C to 175°C; 170°C to 175°C | |
C6H5 NHNHCONHNHC6H5 | |
10 g | |
242.28 | |
Reag. Ph. Eur. |
140-22-7 | |
MFCD00003013 | |
1,5-Diphenylcarbohydrazide; sym.-Diphenylcarbazide | |
≥98% (HPLC); ≥98.0% |
Safety and Handling
RTECSNumber : FF2750000