missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dibromonaphthalene 98.0+%, TCI America™

$85.75 - $378.40

Chemical Identifiers

CAS 83-53-4
Molecular Formula C10H6Br2
Molecular Weight (g/mol) 285.97
MDL Number MFCD00041823
InChI Key IBGUDZMIAZLJNY-UHFFFAOYSA-N
Synonym naphthalene, 1,4-dibromo, 1,4-dibromonapthalene, 1.4-dibromonaphthalene, acmc-209psk, 1,4-dibromo-naphthalen, l ,4-dibromonaphthalene, ksc490o3f, 1,4-dibromonaphthalene 5g
PubChem CID 66521
IUPAC Name 1,4-dibromonaphthalene
SMILES BrC1=C2C=CC=CC2=C(Br)C=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
D23595G
View Documents
TCI America
D23595G
5 g
Each for $85.75
Only null left
 
D235925G
View Documents
TCI America
D235925G
25 g
Each for $378.40
Only null left
 
Description

Description

Specifications

Chemical Identifiers

83-53-4
285.97
IBGUDZMIAZLJNY-UHFFFAOYSA-N
66521
BrC1=C2C=CC=CC2=C(Br)C=C1
C10H6Br2
MFCD00041823
naphthalene, 1,4-dibromo, 1,4-dibromonapthalene, 1.4-dibromonaphthalene, acmc-209psk, 1,4-dibromo-naphthalen, l ,4-dibromonaphthalene, ksc490o3f, 1,4-dibromonaphthalene 5g
1,4-dibromonaphthalene

Specifications

83-53-4
White-Yellow
MFCD00041823
naphthalene, 1,4-dibromo, 1,4-dibromonapthalene, 1.4-dibromonaphthalene, acmc-209psk, 1,4-dibromo-naphthalen, l ,4-dibromonaphthalene, ksc490o3f, 1,4-dibromonaphthalene 5g
BrC1=C2C=CC=CC2=C(Br)C=C1
285.97
285.97
Crystalline Powder
82°C
C10H6Br2
5 g
IBGUDZMIAZLJNY-UHFFFAOYSA-N
1,4-dibromonaphthalene
66521
≥98.0% (GC)
1,4-Dibromonaphthalene
Videos
Safety and Handling

Safety and Handling

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'