missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,4-Diazabicyclo[2.2.2]octane Dihydrobromide 98.0+%, TCI America™
$190.87 - $572.60
Chemical Identifiers
CAS | 54581-69-0 |
---|---|
Molecular Formula | C6H14Br2N2 |
Molecular Weight (g/mol) | 274 |
InChI Key | GPELLQBNVCIHJJ-UHFFFAOYSA-N |
PubChem CID | 68117517 |
IUPAC Name | 1,4-diazabicyclo[2.2.2]octane;dihydrobromide |
SMILES | C1CN2CCN1CC2.Br.Br |
Chemical Identifiers
54581-69-0 | |
274 | |
68117517 | |
C1CN2CCN1CC2.Br.Br |
C6H14Br2N2 | |
GPELLQBNVCIHJJ-UHFFFAOYSA-N | |
1,4-diazabicyclo[2.2.2]octane;dihydrobromide |
Specifications
54581-69-0 | |
White | |
1 g | |
C1CN2CCN1CC2.Br.Br | |
274 | |
274.00 | |
Crystalline Powder |
319°C | |
C6H14Br2N2 | |
GPELLQBNVCIHJJ-UHFFFAOYSA-N | |
1,4-diazabicyclo[2.2.2]octane;dihydrobromide | |
68117517 | |
≥98.0% (N,T) | |
1,4-Diazabicyclo[2.2.2]octane Dihydrobromide |
Safety and Handling
TSCA : No