missing translation for 'onlineSavingsMsg'
Learn More

(1,3-Dioxolan-2-ylmethyl)triphenylphosphonium bromide, 98%

CAS: 52509-14-5 | C22H22BrO2P | 429.29 g/mol

Supplier:  Thermo Scientific Chemicals A1888218

 View more versions of this product

Catalog No. AAA1888218


Only null left
Explore available promotions

Description

Description

It finds its applications as a reagent for Wittig olefinations with introduction of a 1,3-dioxolane moiety. It is applied as a reactant for the preparation of a ratiometric fluorescent probe for specific detection of cysteine over homocysteine and glutathione, for microwave-assisted synthesis of KN-93 as inhibitor of calmodulin kinase II, for the preparation of fluorinated spirobenzofuran piperidines as s1 receptor ligands, in the synthesis and antitumor agents, in regio-selective preparation of indole derivatives via rhodium-catalyzed domino hydroformylation/indolization.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds its applications as a reagent for Wittig olefinations with introduction of a 1,3-dioxolane moiety. It is applied as a reactant for the preparation of a ratiometric fluorescent probe for specific detection of cysteine over homocysteine and glutathione, for microwave-assisted synthesis of KN-93 as inhibitor of calmodulin kinase II, for the preparation of fluorinated spirobenzofuran piperidines as s1 receptor ligands, in the synthesis and antitumor agents, in regio-selective preparation of indole derivatives via rhodium-catalyzed domino hydroformylation/indolization.

Solubility
Soluble in water (20°C).

Notes
Hygroscopic. Store away from water/moisture and oxidizing agents. Store under inert gas. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications

Specifications

(1,3-Dioxolan-2-ylmethyl)triphenylphosphonium bromide
186°C to 189°C
MFCD00011966
Hygroscopic
Soluble in water (20°C).
[Br-].C(C1OCCO1)[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
429.29
429.3
52509-14-5
C22H22BrO2P
50 g
1,3-dioxolan-2-ylmethyl triphenylphosphonium bromide, 1,3-dioxolan-2-yl methyl triphenylphosphonium bromide, 1,3-dioxolan-2-ylmethyl triphenylphosphanium bromide, phosphonium, 1,3-dioxolan-2-ylmethyl triphenyl-, bromide, 1,3-dioxolan-2-yl methyltriphenylphosphonium bromide, 1,3-dioxolan-2-ylmethyl triphenyl phosphanium bromide, 1,3-dioxolan-2-ylmethyl triphenyl phosphonium bromide, 1,3-dioxolan-2-ylmethyl triphenylphosphine, bromide, acmc-209kzy
FRHRVQQUICVJDG-UHFFFAOYSA-M
1,3-dioxolan-2-ylmethyl(triphenyl)phosphanium;bromide
2724194
98%

Chemical Identifiers

52509-14-5
429.29
FRHRVQQUICVJDG-UHFFFAOYSA-M
2724194
[Br-].C(C1OCCO1)[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
C22H22BrO2P
MFCD00011966
1,3-dioxolan-2-ylmethyl triphenylphosphonium bromide, 1,3-dioxolan-2-yl methyl triphenylphosphonium bromide, 1,3-dioxolan-2-ylmethyl triphenylphosphanium bromide, phosphonium, 1,3-dioxolan-2-ylmethyl triphenyl-, bromide, 1,3-dioxolan-2-yl methyltriphenylphosphonium bromide, 1,3-dioxolan-2-ylmethyl triphenyl phosphanium bromide, 1,3-dioxolan-2-ylmethyl triphenyl phosphonium bromide, 1,3-dioxolan-2-ylmethyl triphenylphosphine, bromide, acmc-209kzy
1,3-dioxolan-2-ylmethyl(triphenyl)phosphanium;bromide
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

EINECSNumber : 257-977-3

TSCA : No

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only