missing translation for 'onlineSavingsMsg'
Learn More

1,3,2-Dioxathiolane 2-Oxide 98.0+%, TCI America™

$37.79 - $71.81

Chemical Identifiers

CAS 3741-38-6
Molecular Formula C2H4O3S
Molecular Weight (g/mol) 108.11
MDL Number MFCD00005354
InChI Key WDXYVJKNSMILOQ-UHFFFAOYSA-N
Synonym ethylene sulfite, glycol sulfite, 1,3,2-dioxathiolane, 2-oxide, glycolsulfite, 1,2-ethylene sulfite, cyclic ethylene sulfite, ethylene glycol cyclic sulfite, ethylene glycol, cyclic sulfite, ethylene sulphite, sulfurous acid, cyclic ester with ethylene glycol
PubChem CID 77342
IUPAC Name 1,3,2λ⁴-dioxathiolan-2-one
SMILES O=S1OCCO1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
D29775G
missing translation for 'viewDocuments'
TCI America
D29775G
5 g
Each for $37.79
Only null left
 
D297725G
missing translation for 'viewDocuments'
TCI America
D297725G
25 g
Each for $71.81
Only null left
 
Description

Description

Specifications

Chemical Identifiers

3741-38-6
108.11
WDXYVJKNSMILOQ-UHFFFAOYSA-N
77342
O=S1OCCO1
C2H4O3S
MFCD00005354
ethylene sulfite, glycol sulfite, 1,3,2-dioxathiolane, 2-oxide, glycolsulfite, 1,2-ethylene sulfite, cyclic ethylene sulfite, ethylene glycol cyclic sulfite, ethylene glycol, cyclic sulfite, ethylene sulphite, sulfurous acid, cyclic ester with ethylene glycol
1,3,2λ⁴-dioxathiolan-2-one

Specifications

3741-38-6
172°C
MFCD00005354
ethylene sulfite, glycol sulfite, 1,3,2-dioxathiolane, 2-oxide, glycolsulfite, 1,2-ethylene sulfite, cyclic ethylene sulfite, ethylene glycol cyclic sulfite, ethylene glycol, cyclic sulfite, ethylene sulphite, sulfurous acid, cyclic ester with ethylene glycol
O=S1OCCO1
108.11
108.11
Liquid
Colorless
C2H4O3S
5 g
WDXYVJKNSMILOQ-UHFFFAOYSA-N
1,3,2λ⁴-dioxathiolan-2-one
77342
≥98.0% (GC)
1,3,2-Dioxathiolane 2-Oxide
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (5)-6552

RTECSNumber : KW3900000

TSCA : Yes

SDS
Documents

Documents