missing translation for 'onlineSavingsMsg'
Learn More

1,2-Bis(phenylthio)ethane, 98+%

CAS: 622-20-8 | C14H14S2 | 246.39 g/mol

$109.70 - $336.45

Chemical Identifiers

CAS 622-20-8
Molecular Formula C14H14S2
Molecular Weight (g/mol) 246.39
MDL Number MFCD00014085
InChI Key MHCVYAFXPIMYRD-UHFFFAOYSA-N
Synonym 1,2-bis phenylthio ethane, ethane, 1,2-bis phenylthio, bis phenylthio ethane, 1,1'-ethane-1,2-diylbis thio dibenzene, 2-phenylsulfanyl ethyl sulfanyl benzene, benzene, 1,1'-1,2-ethanediylbis thio bis, 1,1'-1,2-ethanediylbis thio bis-benzene, 1,1'-ethane-1,2-diylbis thio bisbenzene, 2-phenylthioethylthio benzene
PubChem CID 69317
IUPAC Name 2-phenylsulfanylethylsulfanylbenzene
SMILES C(CSC1=CC=CC=C1)SC1=CC=CC=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0201909
View Documents
Thermo Scientific Chemicals
L0201909
10 g
Each for $109.70
Only null left
 
AAL0201918
View Documents
Thermo Scientific Chemicals
L0201918
50 g
Each for $336.45
Only null left
 
Description

Description

1,2-Bis(phenylthio)ethane, is used for the synthesis of the bismaleimide resin, APO-BMI, used in syntactic foams.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2-Bis(phenylthio)ethane, is used for the synthesis of the bismaleimide resin, APO-BMI, used in syntactic foams.

Solubility
Insoluble in water.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Chemical Identifiers

622-20-8
246.39
MHCVYAFXPIMYRD-UHFFFAOYSA-N
69317
C(CSC1=CC=CC=C1)SC1=CC=CC=C1
C14H14S2
MFCD00014085
1,2-bis phenylthio ethane, ethane, 1,2-bis phenylthio, bis phenylthio ethane, 1,1'-ethane-1,2-diylbis thio dibenzene, 2-phenylsulfanyl ethyl sulfanyl benzene, benzene, 1,1'-1,2-ethanediylbis thio bis, 1,1'-1,2-ethanediylbis thio bis-benzene, 1,1'-ethane-1,2-diylbis thio bisbenzene, 2-phenylthioethylthio benzene
2-phenylsulfanylethylsulfanylbenzene

Specifications

622-20-8
C14H14S2
MFCD00014085
1877719
Insoluble in water.
C(CSC1=CC=CC=C1)SC1=CC=CC=C1
246.39
246.39
1,2-Bis(phenylthio)ethane
68°C to 70°C
C6H5SCH2CH2SC6H5
10 g
1,2-bis phenylthio ethane, ethane, 1,2-bis phenylthio, bis phenylthio ethane, 1,1'-ethane-1,2-diylbis thio dibenzene, 2-phenylsulfanyl ethyl sulfanyl benzene, benzene, 1,1'-1,2-ethanediylbis thio bis, 1,1'-1,2-ethanediylbis thio bis-benzene, 1,1'-ethane-1,2-diylbis thio bisbenzene, 2-phenylthioethylthio benzene
MHCVYAFXPIMYRD-UHFFFAOYSA-N
2-phenylsulfanylethylsulfanylbenzene
69317
≥98%
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 210-723-5

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only