missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2-Bis(phenylsulfinyl)ethane Palladium(II) Diacetate 96.0+%, TCI America™
$119.30 - $333.67
Chemical Identifiers
CAS | 858971-43-4 |
---|---|
Molecular Formula | C18H20O6PdS2 |
Molecular Weight (g/mol) | 502.892 |
MDL Number | MFCD09842752 |
InChI Key | SNNYSJNYZJXIFE-UHFFFAOYSA-L |
Synonym | White Catalyst |
PubChem CID | 11562441 |
IUPAC Name | 2-(benzenesulfinyl)ethylsulfinylbenzene;palladium(2+);diacetate |
SMILES | CC(=O)[O-].CC(=O)[O-].C1=CC=C(C=C1)S(=O)CCS(=O)C2=CC=CC=C2.[Pd+2] |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
B3292200MG
|
TCI America
B3292200MG |
200 mg |
Each for $119.30
|
|
|||||
B32921G
|
TCI America
B32921G |
1 g |
Each for $333.67
|
|
|||||
Chemical Identifiers
858971-43-4 | |
502.892 | |
SNNYSJNYZJXIFE-UHFFFAOYSA-L | |
11562441 | |
CC(=O)[O-].CC(=O)[O-].C1=CC=C(C=C1)S(=O)CCS(=O)C2=CC=CC=C2.[Pd+2] |
C18H20O6PdS2 | |
MFCD09842752 | |
White Catalyst | |
2-(benzenesulfinyl)ethylsulfinylbenzene;palladium(2+);diacetate |
Specifications
858971-43-4 | |
MFCD09842752 | |
White Catalyst | |
CC(=O)[O-].CC(=O)[O-].C1=CC=C(C=C1)S(=O)CCS(=O)C2=CC=CC=C2.[Pd+2] | |
502.892 | |
502.89 | |
Crystalline Powder |
C18H20O6PdS2 | |
200 mg | |
SNNYSJNYZJXIFE-UHFFFAOYSA-L | |
2-(benzenesulfinyl)ethylsulfinylbenzene;palladium(2+);diacetate | |
11562441 | |
≥96.0% (T) | |
1,2-Bis(phenylsulfinyl)ethane Palladium(II) Diacetate |
Safety and Handling
TSCA : No
Recommended Storage : Freezer