missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,10-Phenanthroline-5,6-dione 98.0+%, TCI America™
$291.03 - $916.54
Chemical Identifiers
CAS | 27318-90-7 |
---|---|
Molecular Formula | C12H6N2O2 |
Molecular Weight (g/mol) | 210.19 |
MDL Number | MFCD00014473 |
InChI Key | KCALAFIVPCAXJI-UHFFFAOYSA-N |
Synonym | 1,10 phenanthroline-5,6-dione, 1 10-phenanthroline-5 6-dione 97, pyridino 3,2-h quinoline-5,6-dione, phen-5,6-dione, pubchem16755, acmc-209gwd, d0jh7c, phox31, ksc204s8l, 1,2-dione-based compound, 17 |
PubChem CID | 72810 |
IUPAC Name | 5,6-dihydro-1,10-phenanthroline-5,6-dione |
SMILES | O=C1C(=O)C2=CC=CN=C2C2=NC=CC=C12 |
Chemical Identifiers
27318-90-7 | |
210.19 | |
KCALAFIVPCAXJI-UHFFFAOYSA-N | |
72810 | |
O=C1C(=O)C2=CC=CN=C2C2=NC=CC=C12 |
C12H6N2O2 | |
MFCD00014473 | |
1,10 phenanthroline-5,6-dione, 1 10-phenanthroline-5 6-dione 97, pyridino 3,2-h quinoline-5,6-dione, phen-5,6-dione, pubchem16755, acmc-209gwd, d0jh7c, phox31, ksc204s8l, 1,2-dione-based compound, 17 | |
5,6-dihydro-1,10-phenanthroline-5,6-dione |
Specifications
27318-90-7 | |
Yellow | |
MFCD00014473 | |
1,10 phenanthroline-5,6-dione, 1 10-phenanthroline-5 6-dione 97, pyridino 3,2-h quinoline-5,6-dione, phen-5,6-dione, pubchem16755, acmc-209gwd, d0jh7c, phox31, ksc204s8l, 1,2-dione-based compound, 17 | |
O=C1C(=O)C2=CC=CN=C2C2=NC=CC=C12 | |
210.19 | |
210.19 | |
Crystalline Powder |
257°C | |
C12H6N2O2 | |
1 g | |
KCALAFIVPCAXJI-UHFFFAOYSA-N | |
5,6-dihydro-1,10-phenanthroline-5,6-dione | |
72810 | |
≥98.0% (HPLC,T) | |
1,10-Phenanthroline-5,6-dione |
Safety and Handling
TSCA : No