missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,1'-Difluoro-2,2'-bipyridinium Bis(tetrafluoroborate) 95.0+%, TCI America™
$137.95 - $423.30
Chemical Identifiers
CAS | 178439-26-4 |
---|---|
Molecular Formula | C10H8B2F10N2 |
Molecular Weight (g/mol) | 367.792 |
MDL Number | MFCD01073546 |
InChI Key | IDFABAACDRPWQX-UHFFFAOYSA-N |
Synonym | 1,1'-difluoro-2,2'-bipyridinium bis tetrafluoroborate, 1,1'-difluoro-2,2'-bipyridine-1,1'-diium tetrafluoroborate, n,n'-difluoro-2,2'-bipyridinium bis tetrafluoroborate, 1-fluoro-2-1-fluoropyridin-1-ium-2-yl pyridin-1-ium ditetrafluoroborate, mec-31, synfluor reagent, 1,1'-difluoro-2,2'-bipyridine-1,1'-diium ditetrafluoroborate, daikin mec-31, acmc-209eei, n,n'-difluoro-2,2'-bipyridinium |
PubChem CID | 2733931 |
IUPAC Name | 1-fluoro-2-(1-fluoropyridin-1-ium-2-yl)pyridin-1-ium;ditetrafluoroborate |
SMILES | [B-](F)(F)(F)F.[B-](F)(F)(F)F.C1=CC=[N+](C(=C1)C2=CC=CC=[N+]2F)F |
Chemical Identifiers
178439-26-4 | |
367.792 | |
IDFABAACDRPWQX-UHFFFAOYSA-N | |
2733931 | |
[B-](F)(F)(F)F.[B-](F)(F)(F)F.C1=CC=[N+](C(=C1)C2=CC=CC=[N+]2F)F |
C10H8B2F10N2 | |
MFCD01073546 | |
1,1'-difluoro-2,2'-bipyridinium bis tetrafluoroborate, 1,1'-difluoro-2,2'-bipyridine-1,1'-diium tetrafluoroborate, n,n'-difluoro-2,2'-bipyridinium bis tetrafluoroborate, 1-fluoro-2-1-fluoropyridin-1-ium-2-yl pyridin-1-ium ditetrafluoroborate, mec-31, synfluor reagent, 1,1'-difluoro-2,2'-bipyridine-1,1'-diium ditetrafluoroborate, daikin mec-31, acmc-209eei, n,n'-difluoro-2,2'-bipyridinium | |
1-fluoro-2-(1-fluoropyridin-1-ium-2-yl)pyridin-1-ium;ditetrafluoroborate |
Specifications
178439-26-4 | |
C10H8B2F10N2 | |
1 g | |
1,1'-difluoro-2,2'-bipyridinium bis tetrafluoroborate, 1,1'-difluoro-2,2'-bipyridine-1,1'-diium tetrafluoroborate, n,n'-difluoro-2,2'-bipyridinium bis tetrafluoroborate, 1-fluoro-2-1-fluoropyridin-1-ium-2-yl pyridin-1-ium ditetrafluoroborate, mec-31, synfluor reagent, 1,1'-difluoro-2,2'-bipyridine-1,1'-diium ditetrafluoroborate, daikin mec-31, acmc-209eei, n,n'-difluoro-2,2'-bipyridinium | |
[B-](F)(F)(F)F.[B-](F)(F)(F)F.C1=CC=[N+](C(=C1)C2=CC=CC=[N+]2F)F | |
367.792 | |
367.79 | |
Crystalline Powder |
White-Yellow | |
MFCD01073546 | |
1759 | |
IDFABAACDRPWQX-UHFFFAOYSA-N | |
1-fluoro-2-(1-fluoropyridin-1-ium-2-yl)pyridin-1-ium;ditetrafluoroborate | |
2733931 | |
≥95.0% (T) | |
1,1′-Difluoro-2,2′-bipyridinium Bis(tetrafluoroborate) |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator