{ "pagination":{ "page":1, "pageCount":0.8333333333333333, "totalItems":4, "pageSize":12 }, "cartSignIn":"", "ecomm":{ "items":[ {"productRules":{"hasCrossRef":false,"message":"Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com<\/a> for assistance.","suspicious":false,"viewable":true,"buyable":false,"showPrice":false,"noStockOrderDisallow":false,"mustUseMFShipToAddress":false,"partFound":true,"preferred":false,"userType":"G","returnable":true},"priceAndAvailability":[{"availabilityFetched":false,"displayListPrice":false,"showQuantity":false,"hasBreakPrice":false,"whiteListFlag":false,"shippedDirect":false,"displayUnitString":"chaque","uomType":"STD","hasDiscount":false,"validOnRims":false,"hasDynamicPrice":false,"price":"$89,490.04","stockedItem":false,"listPrice":"$89,490.04","uom":"CH","honorWebPrice":false,"quantity":"1","discontinued":false,"unformattedPrice":0.0,"pricingDisplayEU":false,"shippedExpress":false}],"productDetails":{"cdcType":"1","suspicious":false,"vendorRoutingFlag":0,"familyProductUrl":"\/shop\/products\/ibright-fl1500-imaging-system-2\/p-9034006","alternateUnitWidth":0.0,"franchiseNoContent":false,"productDescription":"IBRIGHT FL1500 IMAGING SYSTEM","alternateUnitWeight":0.0,"fisherItem":false,"compliance":"","productCompareEnabled":false,"numStdInAlt":0,"userType":"G","productType":"54","orderInfo":"","vendorPartNumber":"A44241CFR","productFeatures":"
The iBright FL1500 Imaging System supports the main imaging applications of fluorescent, chemiluminescent, and colorimetric western blots, in addition to fluorescent stained nucleic acid gels, fluorescent stained protein gels, colorimetric stained protein gels, and colorimetric membrane stains. We also offer the iBright CL1500 Imaging System, which features the same imaging capabilities as the iBright FL1500 Imaging System, but without fluorescent western blot imaging functionality.<\/p>
Features of the iBright FL1500 Imaging System include:<\/p>