Thermo Scientific SureSTART Sample Handling Solutions
sure-start-sh-guide-d-1-21-2753
Find a Thermo Scientific™ SureSTART™ vial or well plate for nearly every GC or HPLC/UHPLC instrument, application, and budget.
Three distinct SureSTART performance levels — differentiated by instrument compatibility, performance, reproducibility, and value — make selecting the right product simple, clear, and easy.
Why Choose Thermo Scientific SureSTART?
  1. The confidence of knowing every SureSTART product has undergone stringent testing for the desired performance level
  2. Instrument compatibility for all add-on and integrated chromatography autosamplers
  3. Sample security and low compound adsorption that protect valuable samples
  4. Three distinct performance levels to meet your price and performance needs
  5. Extend your consumables budget: The more you buy, the more you save
Find your SureSTART product number with our cross-reference search tool.
Download the SureSTART catalog (PDF, 20.8 MB) to select sample handling products that suit your needs.
Request the Find a SKU/Easy Access Wheel to make finding the right vial simple.
SureSTART Vials and Well Plates: Accurate Results Start Here
Learn more about the three distinct SureSTART performance levels differentiated by instrument compatibility, performance, reproducibility, and value.
Shop By SureSTART Performance Level
Shop NowExplore Performance Level 1Explore Performance Level 2Explore Performance Level 3
 Performance Level Overviews
When to UseCost-optimized products suitable for all HPLC/UHPLC and GC instrument types and tested for everyday analysesHigh-throughput products for high-quality GC and HPLC/UHPLC data with an uninterrupted workflowLowest levels of extractable metals and leachables for consistent and reproducible sensitivity and selectivity, reliable method performance, and sample security and integrity; packaged in a clean environment
Market ExampleAcademic/food analysisFood safety, clinical, environmental, pharma, QC, industrial, forensicsPharma/biopharma, clinical, pharma QC, omics
Instrument TypeVerifying compound presence and quantification using core detector technologies such as UV/Vis, PDA, RI, ELSD, FLD, FID, NPD, ECD, TCD, or FPDRoutine analyses via standard single or triple quadrupole MS systemsHigh-resolution accurate mass (HRAM) technologies, such as Thermo Scientific™ Orbitrap™ mass spectrometer or time of flight (ToF) MS/ion mobility (IMS)
Certification OptionsAutosampler approved — tested to deliver performance, sample security, and reproducibilitySpecification certification by physical parameterCertificate by physical parameters:
  • • LC/GC Certificate using HPLC-UV or GC-FID
  • • MS Certificate using MS
  • • MS Certificate+ using MS, pre-cleaned
Thermo Scientific SureSTART OfferingVials: Glass, plastic, inserts
Caps: AVCS™ caps, snap
Well plates: WebSeal™ plastic
Accessories
Vials: Glass, inserts, headspace, silanized, EPA, specification certified
Caps: AVCS™ caps, bonded, magnetic, snap
Kits: EPA, scintillation, crimpers, de-crimpers
Well plates: WebSeal™ certified plastic
Accessories
Vials: SureStop™ vials, gold-grade, headspace, fixed insert, micro-spring
Caps: AVCS™ caps
Certified Kits: LC/GC, MS CERT, MS CERT+; TOC Electronic crimpers, decrimpers and station
Well plates: WebSeal™ Plate+ (glass-coated)
Accessories
Additional Thermo Scientific Products
Simplifying Selection
Choosing the right vial depends on several set criteria: technique (e.g., LC, GC, or HS), instrument manufacturer, and whether the compounds being detected are light-sensitive or not (do you require clear or amber glass?).
Request our Find a SKU/Easy Access wheel for simplified vial selection when you complete the form.
  • Quick lookup tool helps you find the right vial for your autosampler type
  • One side features recommended part numbers for LC instruments and, the other, GC instruments
Our Fisher Scientific representatives are here to help. Learn more about how you can make your budget go further, or let us be your guide to determine which combination is most suitable for your application requirements.
Contact Us
Complete the form to request our Find a SKU/Easy Access wheel for simplified selection.
* = required field
thermo-fisher-scientific-logo-top-margin
By submitting your data, you agree to our Privacy Policy.
© Thermo Fisher Scientific, Inc. All Rights Reserved.