Solvents Resource Center
solvents-center-resources-banner-d-22-2478
Common Uses of Solvents
Solvents are liquid, gas, or solid chemicals that dissolve, extract, or suspend other substances. The most common and universal solvent is water, which dissolves more substances than any other.
Solvents are often categorized as polar or nonpolar, a factor of the solvent’s structure and charge that determines the types of substances it can dissolve. Polar solvents have a positive and a negative charge at different places in their structures and will dissolve other polar substances. Water, acetone, acetonitrile, dimethylformamide (DMF), dimethyl sulfoxide (DMSO), ethanol, isopropanol, and methanol are examples of polar solvents.
Nonpolar solvents include alkanes (pentanehexane, and heptane) and aromatics (benzene, toluene, and xylene). Other common nonpolar solvents include acetic acid, chloroform, diethyl ether, ethyl acetatemethylene chloride, and pyridine. Nonpolar organic solvents are used as spot removers, paint thinners, nail polish removers, glue solvents, and in perfumes and detergents.
Solvents are used extensively as a medium for chemical reactions and analytical separations and are available in various purity grades and packaging options. They are also essential for manufacturing cosmetics, pharmaceuticals, personal care items, textiles, paints, pesticides, and many other products.
Long-term exposure to some solvents can be hazardous to worker health, however. Chloroform and benzene, for example, are commonly found in gasoline and considered carcinogenic. Solvents can also damage the liver, kidneys, nervous system, or the brain. Chronic solvent-induced encephalopathy (CSE) is a known condition. Occupational exposure to organic solvents is associated with alcoholism, cataracts, and the loss of hearing and color vision.
  More about solvents
molecules-2-21-396-0328
Top Solvents
Learn about each solvent’s physical and chemical properties, uses, designations and grades, and more.
Related Resources
Talk with a Chemical and Chromatography Specialist
Chemical and Chromatography Specialists
Our team is ready to answer your questions and help you select products.
Contact Us
Fisher Scientific Edge
Get Solvents Shipped the Same Day
Find a range of solvents available for same-day shipping through the Fisher Scientific Edge program.
Shop Now
Bulk and Custom Chemical Solutions
Bulk and Custom Chemical Solutions
Our specialists can help you find what works best for your operation.
Contact Us
Chemical Storage and Management Resource Center
Chemical Storage and Management Resource Center
Access safety data sheets, product certificates, and more.
Explore Now
Additional Solvents Resources
Download the following resources to learn more about solvent selection and browse our portfolio.
More
Applications
Chromatography
Chromatography
Find solvents, reagents, and more for liquid and gas chromatography.
Shop Chromatography Products
Cell Analysis Methods
Cell Analysis Methods
Explore products for flow cytometry, cell imaging, and more.
Shop Cell Analysis Products
Protein Biology Methods
Protein Biology Methods
Browse isolation, purification, separation, and detection essentials.
Shop Protein Biology Products
Molecular Biology Methods
Molecular Biology Methods
Explore products and resources for molecular biology protocols.
Shop Molecular Biology Products