Thermo Scientific Temperature Control Products
Accomplish More
We didn't set out to design the world's most innovative baths, circulators, chillers, and coolers. We simply set out to solve the issues that labs like yours face every day — smaller workspaces, tighter budgets, laboratory stock issues, green-building goals and overly complex equipment.
Browse these products:
Temperature Control Selector
An essential tool to help guide the selection of the ideal product for your cooling and heating applications.
Water Baths
Designed to Provide Precise, Reliable Results
Simplify workflows and maximize productivity with Thermo Scientific water baths, featuring enhanced designs. Rugged construction and an advanced temperature controller offer outstanding performance and reliability.
Thermo Scientific™ Precision™ General Purpose Water Baths
Precision general-purpose water baths are rugged, high-performance baths that are designed to maintain water temperature from ambient to 100°C.
Thermo Scientific™ Precision™ Shaking Water Baths
Precision shaking water baths support a range of sensitive life sciences and QA/QC applications, from warming fragile reagents to tissue culturing and genetics sequencing.
Thermo Scientific™ Precision™ Circulating Water Baths
Precision circulating water baths are an ideal choice when temperature uniformity and control are particularly critical, as when working with enzymes or in serological applications.
Thermo Scientific™ Precision™ Coliform Water Baths
Precision coliform water baths, preset to optimal temperatures for easy use, are designed specifically for fecal coliform determination.
Bath and Immersion Circulators
Designed for Optimal Control and Precision
Thermo Scientific Bath Circulators accommodate multiple beakers, test tube racks or incubation vessels with large work areas. Choose from a wide selection of refrigerated bath circulators and heated stainless-steel baths, as well as economical choices. Configure a flexible, cost-effective solution for your application, featuring tight temperature stability and responsive temperature control.
Refrigerated Bath Circulators
Take advantage of outstanding cooling power, expansive temperature ranges, powerful force/suction pumps, and sophisticated digital control technology designed to ensure accuracy and reproducibility of your liquid temperature control procedures. Now offering natural hydrocarbon refrigerants on select models.
Heated Bath Circulators
An economical alternative to stainless steel, these polyphenylene oxide baths are thermally resistant up to 100°C and deliver exceptional temperature performance with operational savings.
Recirculating Chillers
Powerful, Compact Chillers with Full-Range Cooling Capabilities
Support your critical applications that require large process cooling with Thermo Scientific™ recirculating chillers. These reliable solutions offer outstanding performance, value and simplicity. Innovative designs combine flexible configurations with easy-to-use interfaces.
Thermo Scientific Polar Recirculating Chillers
Dynamic, reliable and efficient, the polar series is ideal for closed- and open-looped applications up to 500 watts. Each easy-to-use chiller offers full range cooling for quick ramp-up and cool-down of your application. Designed with you in mind!
Thermo Scientific™ ThermoChill™ Recirculating Chillers
The Thermo Scientific ThermoChill series of chillers is a compact line of refrigerated recirculators offering cooling capacities up to 2000 watts. Innovative design features such as minimized plumbing connections and an integrated full-flow fluid strainer maximize product reliability.
Thermo Scientific™ ThermoFlex™ Recirculating Chillers
Thermo Scientific ThermoFlex recirculating chillers offer a configurable design with a wide range of cooling capacities, an innovative platform that's easy to use, and outstanding performance.
Merlin Recirculating Chillers
The Merlin series of recirculating chillers provide a wide range of cooling capacity all in a reliable, space-saving package for your critical application needs.
Immersion Coolers
Simplify Direct Cooling of Small Samples
Choose a reliable and flexible alternative to tap water cooling with Thermo Scientific immersion coolers. With wide temperature ranges and available integrated control, these immersion coolers are an easy way to cool small samples. Ideal for accessory cooling, or replacing consumables such as dry ice or liquid nitrogen.
Thermo Scientific VersaCool Refrigerated Circulating Bath Flier
Find out how the VersaCool Circulating Bath helps you maximize results and allows for more samples in a small space.
Thermo Scientific VersaCool Refrigerated Circulating Bath Brochure
Learn the benefits of going headless, read FAQs and get specifications and ordering information for the VersaCool.
Thermo Scientific VersaCool Refrigerated Circulating Bath Video
Watch this video to learn about advancements in remote monitoring capabilities, energy efficiency, sample capacity, and other tools and features that’ll give you control.
Precise Temperature Control in Research and Lab Work Video
Detlef Stitterich highlights the importance in achieving precise temperature control in research and lab work.
More
Fisher Scientific Water Savings Flier
Learn about our water savings calculator and how a recirculating chiller or bath circulator can help you lower costs associated with cooling water.
Fisher Scientific Water Savings Calculator Guide
See how much Fisher Scientific's line of circulators and recirculating chillers can save you.
Thermo Scientific VersaCool Refrigerated Circulating Bath 360-Degree Video
Watch a 360 degree viewing of the New! Thermo Scientific VersaCool Refrigerated Circulating 360 Degree Bath.